Invitrogen™

NativeMark™ Unstained Protein Standard

Catalog number: LC0725
Have Questions?
Invitrogen™

NativeMark™ Unstained Protein Standard

Catalog number: LC0725
Have Questions?
Catalog Number
LC0725
Unit Size
5 x 50 µL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
LC07255 x 50 µL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
NativeMark Unstained Protein Standard is a ready-to-use protein standard of eight protein bands in the range of 20–1,200 kDa, designed for molecular weight estimation of proteins using native gel electrophoresis. The protein standard is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use.

Compare and view all other protein standards and ladders ›

Applications
• Sizing estimation of proteins separated on native-polyacrylamide gels and western blots
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Number of Markers
8
Product Type
Protein Ladder
Ready to Load
Yes
Recommended Applications
Native gels
Size Range
20 to 1,200 kDa
Stain Type
Unstained
Gel Compatibility
Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, Novex™Tris-Glycine Gels, NativePAGE™ Gels, NuPAGE™ Tris-Acetate Gels
Molecular Weight (g/mol)
1236, 1048, 720, 480, 242, 146, 66, 20 kDa
Product Line
NativeMARK™
Quantity
5 x 50 μL
Shipping Condition
Dry Ice
System Type
Native-PAGE

Contents & Storage

The NativeMark™ Unstained Protein Standard is supplied in storage buffer (Bis/Tris-HCl, pH 7.0; NaCl; glycerol; Ponceau S).

Store at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text