Novex™ Tricine SDS Buffer Kit, includes LC1676 & LC1675
Novex&trade; Tricine SDS Buffer Kit, <i>includes LC1676 &amp; LC1675</i>
Novex&trade; Tricine SDS Buffer Kit, <i>includes LC1676 &amp; LC1675</i>
Novex&trade; Tricine SDS Buffer Kit, <i>includes LC1676 &amp; LC1675</i>
Novex&trade; Tricine SDS Buffer Kit, <i>includes LC1676 &amp; LC1675</i>
Invitrogen™
Novex™ Tricine SDS Buffer Kit, includes LC1676 & LC1675
The Novex Tricine SDS Buffer Kit is designed for separation of small- to medium-size proteins on tricine gels. This kitRead more
Have Questions?
Catalog NumberQuantity
LC16771 kit
Catalog number LC1677
Price (USD)
172.00
Each
-
Add to cart
Quantity:
1 kit
Price (USD)
172.00
Each
Add to cart
The Novex Tricine SDS Buffer Kit is designed for separation of small- to medium-size proteins on tricine gels. This kit includes the following buffers:
• Tricine SDS Sample Buffer (Cat. No. LC1676)
• Novex Tricine SDS Running Buffer (Cat. No. LC1675)
See all buffers and reagents for SDS-PAGE ›
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Gel TypeSDS PAGE Gel
Quantity1 kit
Gel CompatibilityNovex Tricine Gel
Product LineNovex™
Product TypeBuffer Kit
Unit SizeEach
Contents & Storage
The Novex™ Tricine SDS Buffer Kit includes Novex™ Tricine SDS Running Buffer (10X, 500 ml - LC1675) and Novex™ Tricine SDS Sample Buffer (2X, 20 ml - LC1676).
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link