Nitrocellulose/Filter Paper Sandwich, 0.45 μm, 8.3 x 7.3 cm
Nitrocellulose/Filter Paper Sandwich, 0.45 μm, 8.3 x 7.3 cm
Invitrogen™

Nitrocellulose/Filter Paper Sandwich, 0.45 μm, 8.3 x 7.3 cm

Nitrocellulose/Filter Paper Sandwiches, 0.45 μm, are high-quality membranes ideal for blotting proteins and nucleic acids. The 0.45-μm pore size isRead more
Have Questions?
Catalog NumberQuantity
LC200120 membrane/filter paper sandwiches
Catalog number LC2001
Price (USD)
252.65
Online Exclusive
265.00
Save 12.35 (5%)
Each
-
Add to cart
Quantity:
20 membrane/filter paper sandwiches
Price (USD)
252.65
Online Exclusive
265.00
Save 12.35 (5%)
Each
Add to cart
Nitrocellulose/Filter Paper Sandwiches, 0.45 μm, are high-quality membranes ideal for blotting proteins and nucleic acids. The 0.45-μm pore size is ideal for the transfer of most proteins (>20 kDa) and nucleic acids (>300 bp).

See all membranes and filter papers ›

Features:
Membrane: 100% pure nitrocellulose
Filter paper thickness: 0.8 mm
Binding capacity: ∼80 μg/cm2 of proteins
Compatibility: compatible with commonly used transfer conditions and detection methods such as staining, immunodetection, chemiluminescence, fluorescence, and radiolabeling
• Supplied in a pre-cut, pre-assembled membrane/filter paper sandwich
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionNitrocellulose Pre-Cut Blotting Membrane, 0.45μm pore size
Quantity20 membrane/filter paper sandwiches
Shipping ConditionRoom Temperature
Dimensions (LxW)7.3 cm x 8.3 cm
FormatSandwich
Length (Metric)8.3 cm
MaterialNitrocellulose
Pore Size0.45 μm
Product LineNovex
Thickness0.8 mm
Width (Metric)7.3 cm
Unit SizeEach
Contents & Storage
Store at room temperature.

Frequently asked questions (FAQs)

How should nitrocellulose be treated prior to transferring proteins?

Prewet the nitrocellulose membrane in distilled water or in 1X transfer buffer for 5 min with gentle shaking. This is to prevent any dry spots in the membrane that may interfere with transfer.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

How can I store, strip, and reuse my western blot?

For nitrocellulose or PVDF membrane following Western blot detection using a chemiluminescent or fluorescent substrate system: Following transfer, air dry the membrane and place in an envelope, preferably on top of a supported surface to keep the membrane flat. The blot can be stored indefinitely at -80 degrees C. When ready to reprobe, prewet the PVDF blot with alcohol for a few seconds, followed by a few rinses with pure water to reduce the alcohol concentration. Then proceed as normal with blocking step.

FOR STRIPPING/REPROBING OF MEMBRANES: Harsh protocol (see NOTE below for modifications)

1) Submerge the membrane in stripping buffer (100 mM BME, 2% SDS, 62.5 mM Tris-HCl, pH 6.7) and incubate at 50 degrees C for 30 min with occasional agitation. If more stringent conditions necessary, incubate at 70 degrees C.

2) Wash 2 x 10 min in TBS-T/PBS-T at room temperature.

3) Block the membrane by immersing in 5% blocking reagent TBS-T or PBS-T for 1 hr at room temperature.

4) Immunodetection

NOTE: Often you don't need such harsh conditions to remove antibodies from their proteins. The stringency of one or several of the variables can be decreased: lower the temperature, decrease the time, less BME, less SDS, etc. An especially mild but still often effective stripping protocol is lower pH incubation. Example: pH 2.0 Tris 50-100 mM, 30-60 min incubation (you may do two incubations if you wish). Then rinse and block as usual. If you do not wish to re-use the membrane immediately after stripping, you can store the membrane in plastic wrap (wet, you do not want it to dry out). Another simple, mild stripping buffer is 0.1 M glycine•HCl (pH 2.5-3.0), incubation 30 min to 2 hrs room temperature or 37 degrees C, depending on the antibody.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Are your PVDF and nitrocellulose membranes compatible with the Li-COR instrument?

Yes, both our PVDF and nitrocellulose membranes are compatible with the Li-COR instrument.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link