Nitrocellulose/Filter Paper Sandwiches, 0.2 μm, 8.5 x 13.5 cm
Product Image
Invitrogen™

Nitrocellulose/Filter Paper Sandwiches, 0.2 μm, 8.5 x 13.5 cm

Nitrocellulose/Filter Paper Sandwiches, 0.2 μm, are high-quality membranes ideal for blotting proteins and nucleic acids. The 0.2-μm pore size isRead more
Have Questions?
Catalog NumberQuantity
LC200916 membrane/filter paper sandwiches
Catalog number LC2009
Price (USD)
298.65
Online Exclusive
313.00
Save 14.35 (5%)
Each
In stock
Add to cart
Quantity:
16 membrane/filter paper sandwiches
Recurring order eligible. Learn more »
Price (USD)
298.65
Online Exclusive
313.00
Save 14.35 (5%)
Each
Add to cart
Ask our AI about this Product
Nitrocellulose/Filter Paper Sandwiches, 0.2 μm, are high-quality membranes ideal for blotting proteins and nucleic acids. The 0.2-μm pore size is ideal for the transfer of low molecular weight proteins (<20 kDa) or peptides and nucleic acids (<300 bp). The included extra-thick, absorbent blotting paper (2.5 mm) accommodates blotting set-up using semi-dry transfer apparatuses.

See all membranes and filter papers ›

Features:
Membrane: 100% pure nitrocellulose
Filter paper thickness: 2.5 mm
Binding capacity: ∼80 μg/cm2 of proteins
Binding interactions: hydrophobic and electrostatic
Compatibility: compatible with commonly used transfer conditions and detection methods such as staining, immunodetection, chemiluminescence, fluorescence, and radiolabeling
• Supplied in a pre-cut, pre-assembled membrane/filter paper sandwich
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionNitrocellulose/Filter Paper Sandwich, 0.2μm pore size
Quantity16 membrane/filter paper sandwiches
Shipping ConditionRoom Temperature
Dimensions (LxW)8.5 cm x 13.5 cm
FormatSandwich
Length (Metric)13.5 cm
MaterialNitrocellulose
Pore Size0.2 μm
Product LineNovex
Thickness2.5 mm
Width (Metric)8.5 cm
Unit SizeEach
Contents & Storage
Store at room temperature.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

What types of membrane work best with SuperSignal West Pico PLUS Chemiluminescent Substrate?

Most researchers use nitrocellulose or polyvinyldiflouride (PVDF) membranes with SuperSignal West Dura Chemiluminescent Substrate. Both work well, although nitrocellulose seems to be better suited in some applications than PVDF. In addition, charge-modified nylon membrane performs well with this substrate. Please also see our guide for choosing western blot membranes (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/western-blotting/transfer-proteins-western-blot/membranes-transfer-buffers-western-blotting/membranes-western-blotting.html).

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

What types of membrane work best with SuperSignal West Femto Maximum Sensitivity Substrate?

Most researchers use nitrocellulose or polyvinyldiflouride (PVDF) membranes with SuperSignal West Femto Maximum Sensitivity Substrate. Both work well, although nitrocellulose seems to be better suited in some applications than PVDF. In addition, charge-modified nylon membrane performs well with this substrate. Please also see our guide for choosing western blot membranes (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/western-blotting/transfer-proteins-western-blot/membranes-transfer-buffers-western-blotting/membranes-western-blotting.html).

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

What types of membrane work best with SuperSignal West Dura Extended Duration Substrate?

Most researchers use nitrocellulose or polyvinyldiflouride (PVDF) membranes with SuperSignal West Dura Extended Duration Substrate. Both work well, although nitrocellulose seems to be better suited in some applications than PVDF. In addition, charge-modified nylon membrane performs well with this substrate. Please also see our guide for choosing western blot membranes (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/western-blotting/transfer-proteins-western-blot/membranes-transfer-buffers-western-blotting/membranes-western-blotting.html).

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

How should nitrocellulose be treated prior to transferring proteins?

Prewet the nitrocellulose membrane in distilled water or in 1X transfer buffer for 5 min with gentle shaking. This is to prevent any dry spots in the membrane that may interfere with transfer.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

How can I store, strip, and reuse my western blot?

For nitrocellulose or PVDF membrane following Western blot detection using a chemiluminescent or fluorescent substrate system: Following transfer, air dry the membrane and place in an envelope, preferably on top of a supported surface to keep the membrane flat. The blot can be stored indefinitely at -80 degrees C. When ready to reprobe, prewet the PVDF blot with alcohol for a few seconds, followed by a few rinses with pure water to reduce the alcohol concentration. Then proceed as normal with blocking step.

FOR STRIPPING/REPROBING OF MEMBRANES: Harsh protocol (see NOTE below for modifications)

1) Submerge the membrane in stripping buffer (100 mM BME, 2% SDS, 62.5 mM Tris-HCl, pH 6.7) and incubate at 50 degrees C for 30 min with occasional agitation. If more stringent conditions necessary, incubate at 70 degrees C.

2) Wash 2 x 10 min in TBS-T/PBS-T at room temperature.

3) Block the membrane by immersing in 5% blocking reagent TBS-T or PBS-T for 1 hr at room temperature.

4) Immunodetection

NOTE: Often you don't need such harsh conditions to remove antibodies from their proteins. The stringency of one or several of the variables can be decreased: lower the temperature, decrease the time, less BME, less SDS, etc. An especially mild but still often effective stripping protocol is lower pH incubation. Example: pH 2.0 Tris 50-100 mM, 30-60 min incubation (you may do two incubations if you wish). Then rinse and block as usual. If you do not wish to re-use the membrane immediately after stripping, you can store the membrane in plastic wrap (wet, you do not want it to dry out). Another simple, mild stripping buffer is 0.1 M glycine•HCl (pH 2.5-3.0), incubation 30 min to 2 hrs room temperature or 37 degrees C, depending on the antibody.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3062998Certificate of AnalysisMar 27, 2025LC2009
2983000Certificate of AnalysisSep 09, 2024LC2009
2868480Certificate of AnalysisJun 03, 2024LC2009
2725110Certificate of AnalysisApr 16, 2024LC2009
2725109Certificate of AnalysisAug 04, 2023LC2009
5 results displayed, search above for a specific certificate

Safety Data Sheets

Other products to consider



Share catalog number, name or link