Invitrogen™

Fluorescent Compatible Sample Buffer (4X, non-reducing)

Catalog number: LC2570
Have Questions?
Invitrogen™

Fluorescent Compatible Sample Buffer (4X, non-reducing)

Catalog number: LC2570
Have Questions?
Catalog Number
LC2570
Unit Size
5 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
LC25705 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Invitrogen Fluorescent Compatible Sample Buffer is for use in SDS-polyacrylamide gel electrophoresis (SDS-PAGE). It is optimized for use in fluorescent western blotting as it does not interfere in fluorescent channels, unlike other standard sample buffers. Fluorescent Compatible Sample Buffer is non-reducing but may also be used under denaturing conditions.

Non-reducing—ready-to-use for non-reducing SDS-PAGE, or the preferred type and amount of reducing agent (e.g., DTT) can be added to produce reducing conditions
Lane marker dyes—visible markers of the dye front
Convenient—stable at room temperature, enabling storage at the bench where electrophoresis is performed
Concentrated—4X formulation provides more versatility than traditional 2X buffers
Ideal for fluorescent detection—formulated to prevent auto fluorescence during fluorescent detection

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Buffer
Sample Loading Buffers
Concentration
4X
Detection Method
Fluorescent
Gel Type
Tricine, Tris-Glycine, SDS-PAGE, Bis-Tris, Bis-Tris Plus, Tris-Acetate
Product Type
Fluorescent Compatible Sample Buffer
Gel Compatibility
SDS-PAGE Gels
Shipping Condition
Room Temperature

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text