Invitrogen™

Novex™ Tris-Glycine Native Sample Buffer (2X)

Catalog number: LC2673
Have Questions?
Invitrogen™

Novex™ Tris-Glycine Native Sample Buffer (2X)

Catalog number: LC2673
Have Questions?
Catalog Number
LC2673
Unit Size
20 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
LC267320 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Novex Tris-Glycine Native Sample Buffer (2X) is designed for protein gel electrophoresis under native running conditions with Tris-Glycine gels or Tris-Acetate gels. It has a pH of 8.6 and contains bromophenol blue as a tracking dye.

See all available buffers and reagents available for Native-PAGE

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Name or Material
Sample Loading Buffer
Recommended Storage
Store in refrigerator (2–8°C).
Concentration
2X
For Use With (Equipment)
Mini Precast Protein Gels
pH
8.6
Physical Form
Liquid
Product Line
Novex
Quantity
20 mL
Formulation
Glycerol 56-81-5 (20-30)

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text