Novex™ Tris-Glycine SDS Running Buffer (10X)
Novex™ Tris-Glycine SDS Running Buffer (10X)
Invitrogen™

Novex™ Tris-Glycine SDS Running Buffer (10X)

Novex Tris-Glycine SDS Running Buffer (10X) is formulated for separation of proteins in their denatured state on Tris-Glycine gels. Tris-GlycineRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
LC2675500 mL
LC267544 x 1 L
LC267555 L
Catalog number LC2675
Price (USD)
38.65
Online Exclusive
39.91
Save 1.26 (3%)
Each
In stock
Add to cart
Quantity:
500 mL
Price (USD)
38.65
Online Exclusive
39.91
Save 1.26 (3%)
Each
Add to cart
Novex Tris-Glycine SDS Running Buffer (10X) is formulated for separation of proteins in their denatured state on Tris-Glycine gels. Tris-Glycine gels provide reproducible separation of a wide range of proteins into well-resolved bands.

Separate native or denatured proteins
Novex Tris-Glycine gels do not contain SDS and can be used to accurately separate both native and denatured proteins, depending upon the sample and running buffers used. To separate denatured proteins on Novex Tris-Glycine gels, use Novex Tris-Glycine SDS Sample Buffer and Novex Tris-Glycine SDS Running Buffer. To separate native proteins use Novex Tris-Glycine Native Sample Buffer and Novex Tris-Glycine Native Running Buffer.

See all available buffers and reagents available for SDS-PAGE

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialRunning Buffer
Recommended StorageContains: Novex™ Tris-Glycine SDS Running Buffer (10X), 500 ml
Storage: +4°C to 25°C

Guaranteed stable for 6 months unless otherwise specified in the product literature.
Concentration10X
Physical FormLiquid
Product LineNovex
Quantity500 mL
Unit SizeEach

Frequently asked questions (FAQs)

Where do I find buffer recipes for your precast protein gels?

The formulations of buffers for our precast protein gels can be found at this link: https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-gel-electrophoresis/protein-electrophoresis-buffers-reagents.html

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

If a Tricine gel is accidentally run with buffers used in the Tris-Glycine system, what will happen and why?

If the Tricine gel is run with Tris-Glycine sample buffer, the bands will behave abnormally and resolve poorly. If the Tricine gel is accidentally run with Tris-Glycine running buffer, the gel will take longer to run and the resolution, especially for smaller proteins, will be worse than when the proteins are run on a Tris-Glycine gel with Tris-Glycine buffers. This is due to a combination of increase in stack area size (glycine is a slower ion than Tricine) and the higher ionic strength of the Tricine gel.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

How should I store Novex Tris-Glycine SDS Running Buffer (10X)?

We recommend storing Novex Tris-Glycine SDS Running Buffer (10X) at 4 to 25 degrees C.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

What are the recommended sample loading volumes and protein loading amounts for your precast protein gels?

*Tris-Glycine and Invitrogen Tricine Mini gels: see here (http://tools.thermofisher.com/content/sfs/manuals/electrophoresisguide_man.pdf), Page 8

*NuPAGE Tris-Acetate and NuPAGE Bis-Tris Mini gels: see here (http://tools.thermofisher.com/content/sfs/manuals/nupage_tech_man.pdf), Page 10

*Bolt Bis-Tris Plus Mini gels: see here (http://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-gel-electrophoresis/protein-gels/bolt-bis-tris-gels.html)

*Thermo Scientific Precise Tris-HEPES gels: see here (https://tools.thermofisher.com/content/sfs/manuals/MAN0011499_Precise_Protein_Gels_UG.pdf), Page 1

*Midi gels (Invitrogen Tris-Glycine, NuPAGE Bis-Tris and NuPAGE Tris-Acetate): see here (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/novex_midigel_man.pdf), Page 4

*Thermo Scientific Precise Tris-Glycine gels: see here (https://tools.thermofisher.com/content/sfs/manuals/D25MAN0011814_Precise_TrisGlycine_Gels_UG.pdf), Page 1

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Do your precast protein gels contain SDS?

Our precast protein gels do not contain SDS but they can be run under denaturing conditions when used with the appropriate denaturing running buffer.
Note: NuPAGE Bis-Tris gels, Bolt Bis-Tris Plus gels, and Thermo Scientific Precise Tris-HEPES gels cannot be run under native conditions; they can only be run under denaturing conditions.

*Invitrogen Tris-Glycine gels: For Native electrophoresis, use Invitrogen Tris-Glycine Native Running Buffer. For Denaturing electrophoresis, use Invitrogen Tris-Glycine SDS Running Buffer

*NuPAGE Tris-Acetate gels: For Native electrophoresis, use Invitrogen Tris-Glycine Native Running Buffer. For Denaturing electrophoresis, use NuPAGE Tris-Acetate SDS Running Buffer

*NuPAGE Bis-Tris gels: For Denaturing electrophoresis, use NuPAGE MOPS-SDS Running Buffer or NuPAGE MES-SDS Running Buffer

*Bolt Bis-Tris Plus gels: For Denaturing electrophoresis, use Bolt MOPS SDS Running Buffer or Bolt MES SDS Running Buffer

*Thermo Scientific Precise Tris-Glycine gels: For Native electrophoresis, use Tris-Glycine SDS Running Buffer without SDS added. For Denaturing electrophoresis, use Tris-Glycine SDS Running Buffer.

*Thermo Scientific Precise Tris-HEPES gels: For Denaturing electrophoresis, use Tris-HEPES SDS Running Buffer.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link