Invitrogen™

Tris-Glycine Transfer Buffer (25X)

Catalog number: LC3675
Have Questions?
Invitrogen™

Tris-Glycine Transfer Buffer (25X)

Catalog number: LC3675
Have Questions?
Catalog Number
LC3675
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
LC3675500 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Tris-Glycine Transfer Buffer (25X) is optimized for western blot transfer of proteins to nitrocellulose, PVDF, or nylon membranes for subsequent analysis using Tris-Glycine gels.

To use: This buffer should be diluted to a 1X solution with a water/methanol mixture to yield a final methanol concentration of 20% for optimal results.

See all available buffers and reagents available for SDS-PAGE

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
25X
Gel Type
Tris-Glycine Gel
Shipping Condition
Room Temperature
Gel Compatibility
Novex Tris-Glycine Gel
Membrane Compatibility
Nitrocellulose, PVDF, Nylon
Product Line
Novex™
Product Type
Transfer Buffer

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text