MagicMark™ XP Western Protein Standard
MagicMark™ XP Western Protein Standard
Invitrogen™

MagicMark™ XP Western Protein Standard

MagicMark XP Western Protein Standard consists of nine recombinant proteins (20–220 kDa), each of which contains an IgG binding site.Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
LC560350 μL
LC5602250 μL
LC5602X44 x 250 μL
3 Options
Catalog number LC5603
Price (USD)/ 50 µL
133.65
Online exclusive
139.00 
Save 5.35 (4%)
-
Add to cart
Quantity:
50 μL
Request bulk or custom format
Price (USD)/ 50 µL
133.65
Online exclusive
139.00 
Save 5.35 (4%)
Add to cart
MagicMark™ XP Western Protein Standard
Catalog numberLC5603
Price (USD)/ 50 µL
133.65
Online exclusive
139.00 
Save 5.35 (4%)
-
Add to cart
MagicMark XP Western Protein Standard consists of nine recombinant proteins (20–220 kDa), each of which contains an IgG binding site. The IgG binding site binds the primary or secondary antibody used for detection of the target protein, allowing direct visualization of the standard on the western blot. The MagicMark XP Standard is compatible with most western kits and substrates (chemiluminescent, chromogenic, and fluorescent). The protein standard is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use.

Compare and view all other protein standards and ladders ›

Applications
• Western blotting: detection of the nine unstained bands via the detection method used for the target protein. Compatible with chemiluminescent substrates and fluorescent secondary antibodies (not recommended for antibodies labeled with fluors in the 500–550 nm channel).
• Protein size estimation: protein sizing on SDS-polyacrylamide gels and on western blot using western blot detection method
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodUser defined detection system
Number of Markers9
Product TypeProtein Ladder
Ready to LoadYes
Size Range20 to 220 kDa
Stain TypeUnstained
Gel CompatibilityNovex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels
Molecular Weight (g/mol)220, 120, 100, 80, 60, 50, 40, 30, 20 kDa
Product LineMagicMark™
Quantity50 μL
Shipping ConditionDry Ice
System TypeWestern Blotting, SDS-PAGE
Unit Size50 µL
Contents & Storage
50 μL of ready-to-use standard mixture is provided in a plastic vial. Loading buffer consists of 125 mM Tris-HCl (pH 6.8), 10 mM DTT, 17.4% Glycerol, 3% SDS, and 0.025% Bromophenol Blue.

Store at -20°C.