Invitrogen™

Novex™ Sharp Pre-stained Protein Standard

Catalog number: LC5800
Have Questions?
Invitrogen™

Novex™ Sharp Pre-stained Protein Standard

Catalog number: LC5800
Have Questions?
Catalog Number
LC5800
Unit Size
2 x 250 µL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
LC58002 x 250 µL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Novex Sharp Pre-Stained Protein Standard is designed for accurate, easy, and convenient molecular weight estimation of a wide range of molecular weight proteins during SDS-PAGE and western blotting. The standard consists of 12 colored bands in the range of 3.5 to 260 kDa and is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use.

Compare and view all other protein standards and ladders ›

Applications
• Monitoring protein migration during SDS-polyacrylamide gel electrophoresis
• Monitoring protein transfer onto membranes after western blotting
• Sizing of proteins on SDS-PAGE gels and western blots

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Colorimetric
Number of Markers
12
Product Type
Protein Ladder
Ready to Load
Yes
Size Range
3.5 to 260 kDa
Stain Type
3 colors: Pink, Yellow, Blue
Gel Compatibility
Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels
Molecular Weight (g/mol)
260, 160, 110, 80, 60, 50, 40, 30, 20, 15, 10, 3.5 kDa
Product Line
Novex™
Quantity
2 x 250 μL
Shipping Condition
Approved for shipment on Wet or Dry Ice
System Type
Western Blotting, SDS-PAGE

Contents & Storage

Novex™ Sharp Pre-stained Protein Standards are provided as 2 x 250 μL (total of 50 applications of 10 μL each) of ready-to-use standard mixture. The Novex™ Sharp Pre-Stained standard loading buffer consists of 65 mM Tris pH 6.5, 30% glycerol, 2% SDS, and 2.5 mM ETDA.

Store at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text