Invitrogen™

Colloidal Blue Staining Kit

Catalog number: LC6025
Have Questions?
Invitrogen™

Colloidal Blue Staining Kit

Catalog number: LC6025
Have Questions?
Catalog Number
LC6025
Unit Size
1 kit*
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
LC60251 kit*
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Colloidal Blue Staining Kit allows detection of nanogram levels of proteins in 1D or 2D PAGE gels with minimal effort and requires only water to destain. The Colloidal Blue stain uses colloidal chemistry that reduces free dye in solution and improves the protein-to-dye binding ratio. Samples are intensely stained and visible within three hours. Background staining is virtually eliminated by destaining overnight with water. The kit requires one easy solution preparation. Methanol is required in the staining step, but is not included in the kit.

Compare all Coomassie stains ›
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Location
In-Gel Detection
Detection Method
Colorimetric
Label or Dye
Coomassie
Product Type
Protein Gel Staining Kit
Shelf Life
6 Months
Shipping Condition
Room Temperature
Target Molecule
Protein

Contents & Storage

The Colloidal Blue Staining Kit is supplied with sufficient stainer A and stainer B to stain 25 mini-gels. Store the kit at room temperature. The kit is guaranteed for 6 months when properly stored unless otherwise stated in the product documentation.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text