Invitrogen™

InVision™ His-Tag In-Gel Staining Kit

Catalog number: LC6033
Have Questions?
Invitrogen™

InVision™ His-Tag In-Gel Staining Kit

Catalog number: LC6033
Have Questions?
Catalog Number
LC6033
Unit Size
1 kit*
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
LC60331 kit*
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The InVision His-tag In-gel Staining Kit includes InVision His-tag In-gel Stain as well as BenchMark His-tagged Protein Standard. InVision His-tag In-gel Stain is a ready-to-use, fast, sensitive, and highly specific stain for visualizing His-tagged fusion protein bands directly in a polyacrylamide gel following electrophoresis.

Features:
• Stain proteins fused to an oligohistidine sequence
• Eliminate western blotting—detect His-tagged proteins directly in the gel
• Detect nanogram levels of protein bands with a UV transilluminator or imaging equipment with appropriate filters or lasers
• Visualize results in under three hours

Compare all His-tag stains ›

Fast and easy staining
The InVision His-tag In-gel Stain protocol is easy to perform and requires very little hands-on-time, allowing for rapid and scalable protein expression screening in a variety of gel types. Following electrophoresis, the gel is fixed, incubated in the stain, washed, and the specific His-tagged fusion protein detected by exposing the gel to UV or visible light. In under three hours, nanogram levels of His-tagged fusion protein can be detected. After documenting your InVision Stain signal, total protein staining can be performed.
WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Location
In-Gel Detection
Detection Method
Fluorescent
Label or Dye
InVision His-Tag Stain
Product Type
His Tag Proteins In-Gel Staining Kit
Product Line
InVision™
Quantity
1 kit
Target Molecule
Proteins (His-Tagged)

Contents & Storage

InVision™ His-tag In-gel Stain is supplied as a 1X ready-to-use staining reagent (500 mL). BenchMark™ His-tagged Protein Standard (125 μL) is also included in the kit.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text