Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | SimplyBlue™ SafeStain, 3.5 L |
Detection Location | In-Gel Detection |
Detection Method | Colorimetric |
Label or Dye | Coomassie |
Product Type | Safe Protein Stain |
Shelf Life | 6 Months |
Green Features | Less hazardous |
Product Line | SimplyBlue™ |
Quantity | 3.5 L |
Target Molecule | Protein |
Unit Size | 3.5 L |
Description | SimplyBlue™ SafeStain, 1 L |
Detection Location | In-Gel Detection |
Detection Method | Colorimetric |
Label or Dye | Coomassie |
Product Type | Safe Protein Stain |
Shelf Life | 6 Months |
Green Features | Less hazardous |
Product Line | SimplyBlue™ |
Quantity | 1 L |
Target Molecule | Protein |
Unit Size | 1 L |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
LC6065 | 3.5 L | SimplyBlue™ SafeStain, 3.5 L | 467.65 Online price 506.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
LC6060 | 1 L | SimplyBlue™ SafeStain, 1 L | 199.65 Online price 213.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|