Invitrogen™

SilverQuest™ Silver Staining Kit

Catalog number: LC6070
Have Questions?
Invitrogen™

SilverQuest™ Silver Staining Kit

Catalog number: LC6070
Have Questions?
Catalog Number
LC6070
Unit Size
1 kit*
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
LC60701 kit*
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The SilverQuest Silver Staining Kit provides a rapid and easy method to silver stain proteins and DNA in polyacrylamide gels. It is specifically designed to provide sensitive silver staining compatible with mass spectrometry analysis.

Compare all silver stains ›

Features of this kit:
Compatible—staining compatible with mass spectrometry analysis
Protocol—protein bands (0.3 ng of protein) can be visualized within an hour or for added speed a microwave procedure can be completed in ∼30 minutes
Color-coded solutions—color-coded solutions for easy staining protocol

Mass spectrometry-compatible reagents
The SilverQuest Silver Staining Kit has been specifically designed to make protein detection compatible with mass spectrometry analysis. The SilverQuest reagent set contains a sensitizer that contains no glutaraldehyde or formaldehyde, so it leaves lysine and arginine side-chains unmodified. Additionally, the SilverQuest destaining solution effectively removes silver from protein bands in polyacrylamide gels. Peptides remain as efficient targets for in-gel trypsin digestion and gel extraction and yield better coverage during mass spectrometry analysis.
WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Location
In-Gel Detection
Detection Method
Colorimetric
Label or Dye
SilverQuest Silver Stain
Product Type
Protein Gel Staining Kit
Shelf Life
6 Months
Product Line
SilverQuest™
Target Molecule
Protein

Contents & Storage

This kit contains sufficient reagents to stain 25 minigels

Contains:
250 mL Sensitizer
25 mL Stainer
250 mL Developer
2 mL Developer Enhancer
250 mL Stopper solution
60 mL Destainer A
60 mL Destainer B

Store kit at room temperature (15°C to 30°C). Stable for 6 months.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text