Invitrogen™

Lumio™ Green Detection Kit

Catalog number: LC6090
Have Questions?
Invitrogen™

Lumio™ Green Detection Kit

Catalog number: LC6090
Have Questions?
Catalog Number
LC6090
Unit Size
1 kit*
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
LC60901 kit*
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Lumio™ Green Detection Kit is a sensitive and highly specific kit for labeling Lumio™ fusion proteins prior to electrophoresis. The Lumio™ Green Kit enables immediate visualization of Lumio™ fusion protein bands directly in a polyacrylamide gel. Using the Lumio™ Green Detection Kit you'll:

• Label proteins fused to a Lumio™ sequence
• Visualize results immediately after electrophoresis—eliminate gel staining and western blotting
• Detect nanogram levels of protein with a UV transilluminator, equipped with a standard camera (Figure 1), or a laser-based scanner
The Lumio™ Green Detection Reagent, included in the Lumio™ Green Detection Kit, can also be used for real-time detection of protein synthesis during in vitro transcription-translation reactions.

Immediate and Effortless Detection
To detect using the Lumio™ Green Detection Kit simply:
1. Add the Lumio™ Green Detection Reagent and the optimized Lumio™ Sample Buffer to your protein sample and heat for 10 minutes at 70°C.
2. Add the Lumio™ In-Gel Detection Enhancer to your sample mix and incubate for 5 minutes at room temperature.
3. Load and electrophorese your protein gel.
4. Visualize your Lumio™ fusion protein bands with a UV transilluminator, equipped with a standard camera, or laser-based scanner.
5. After documenting your Lumio™ Green signal, total protein staining can be performed.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Location
In-Gel Detection
Detection Method
Fluorescent
Label or Dye
Lumio Green
Product Type
Pre-Gel Fusion Protein Labeling Kit
Shelf Life
6 Months
Product Line
Lumio™
Shipping Condition
Dry Ice
Target Molecule
Proteins (Lumio™-Tagged) , Proteins (TC-Tagged)

Contents & Storage

The Lumio™ Green Detection Kit contains one vial of Lumio™ Green Detection Reagent, five vials of Lumio™ Gel Sample Buffer, and one vial of Lumio™ In-Gel Detection Enhancer. Store at -20°C. All components are guaranteed stable for 6 months when properly stored.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text