CapSure™ Macro LCM Caps
CapSure™ Macro LCM Caps
Applied Biosystems™
CapSure™ Macro LCM Caps
Specially designed for use with the Arcturus™ Laser Capture Microdissection (LCM) instruments, CapSure™ HS and CapSure Macro LCM Caps enableRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
LCM0212240 Caps
LCM021148 Caps
Catalog number LCM0212
Price (USD)
2,835.00
Each
-
Add to cart
Quantity:
240 Caps
Price (USD)
2,835.00
Each
Add to cart
Ask our AI about this Product
Specially designed for use with the Arcturus™ Laser Capture Microdissection (LCM) instruments, CapSure™ HS and CapSure Macro LCM Caps enable the precise and rapid extraction of pure populations of cells from heterogeneous sample preparations. CapSure Caps may be used to isolate individual cells or thousands of cells on a single cap.
Key product features:
• Gentle application – non-damaging microdissection preserves the integrity of the captured material
• Versatile extraction – capture both large and small amounts of cells
Ensure the Integrity of Microdissected Cells for Downstream Molecular Analysis
All CapSure™ LCM Caps have a patented transfer film bonded to the lower cap surface. Using the Arcturus Microdissection Systems an infrared laser pulses through the top of the cap during LCM and interacts with the transfer film, which then melts and bonds to the cells or regions of interest. The film absorbs the laser radiation – instead of the tissue or cell sample – creating a gentle, non-damaging microdissection that preserves the integrity of the captured material.
Extract Large Amounts of Cells with CapSure™ Macro LCM Caps
CapSure™ Macro LCM Caps provide the maximum area for capturing several thousand cells on a single cap. This is particularly useful for downstream applications that require large numbers of cells to obtain enough material for analysis, such as some protein analysis techniques. The CapSure™ Macro LCM Caps fit directly onto 0.5 ml microcentrifuge tubes for extraction of biological molecules from captured cells.
Extract Small Numbers of Cells with High Sensitivity CapSure™ HS Caps
CapSure™ HS LCM Caps enable the highly sensitive extraction and detection of biological molecules from small numbers of cells. CapSure™ HS LCM Caps contain ridges at the bottom that keep the active capture area of the cap twelve microns off the surface of the sample. This separation ensures the precise capture of up to several hundreds of cells. In addition, the CapSure™ LCM Caps work with ExtracSure devices that minimize the dilution of biological molecules extracted from captured cells. This optimizes the recovery of these molecules for downstream molecular analysis.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)ArcturusXT™ LCM Instrument
IncludesCapsure Macro LCM Caps
MaterialLCM
Quantity240 Caps
Product LineCapSure
TypeCap
Unit SizeEach
Contents & Storage
CapSure Macro LCM Caps. Store at room temperature
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Frequently asked questions (FAQs)
Are CapSure LCM Caps compatible with instruments other than ArcturusXT LCM instruments?
CapSure LCM Caps are specifically designed for use with Arcturus LCM instruments. Therefore, we do not recommend using them with other instruments.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection