Applied Biosystems™

Incubation Block for Sample Extraction from CapSure™ HS Caps

Catalog number: LCM0505
Have Questions?
Applied Biosystems™

Incubation Block for Sample Extraction from CapSure™ HS Caps

Catalog number: LCM0505
Have Questions?
Catalog Number
LCM0505
Unit Size
1 each
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
LCM05051 each
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Use this accessory for the ARCTURUS™ Laser Capture Microdissection (LCM) instrument to ensure success in your microgenomics research.

Alignment Tray and Incubation Block
The Alignment Tray ensures alignment of the CapSure™ HS LCM Cap and ExtracSure™ Device during the extraction process, while the Incubation Block keeps your sample at a constant temperature. Used together, they maximize the efficiency of extraction of biomolecules from microdissected cells on captured on CapSure™ HS LCM caps.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Type
Incubation Block
Product Line
CapSure
For Use With (Equipment)
ArcturusXT™ LCM Instrument
Includes
Incubation Block
Shipping Condition
Room Temperature

Contents & Storage

Incubation block

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text