Applied Biosystems™

PEN Membrane Frame Slides for Live Cell Microdissection

Catalog number: LCM0530
Have Questions?
Applied Biosystems™

PEN Membrane Frame Slides for Live Cell Microdissection

Catalog number: LCM0530
Have Questions?
Catalog Number
LCM0530
Unit Size
5 each
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
25 each
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
LCM05305 each
-
LCM053125 each
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

A suite of accessories has been designed to enable research on living cells using your ARCTURUSXT™ LCM Instrument. Researchers may use slides or Petri dishes to perform imaging and microdissection experiments. Following a validated protocol from ARCTURUS™ LCM Systems from Applied Biosystems™mdash;the leaders in tissue-based microdissection—the live cells may be isolated, remaining viable for subsequent re-culture or direct processing for use in downstream applications.

Polyethylene naphthalate (PEN) Membrane Frame Slides for Live Cell Microdissection
• Specially optimized for use with live cell experiments
• Take advantage of the CapSure™ LCM Cap technology to collect cells for downstream processing

For Research Use Only. Not for use in diagnostics procedures.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Type
PEN Membrane Frame Slide
For Use With (Equipment)
ArcturusXT™ LCM Instrument
Includes
Live Cell PEN Membrane Frame Slides
Material
Polyethylene Naphthalate
Quantity
5
Shipping Condition
Room Temperature

Contents & Storage

Live Cell PEN Membrane Frame Slides. Store at room temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text