LCQUAN™ Quantitative Software
An important software update is available for all Thermo Scientific Xcalibur/Foundation software users. Download now.
LCQUAN™ Quantitative Software
Thermo Scientific™

LCQUAN™ Quantitative Software

Acquire and manage quantitative LC-MS data in a 21 CFR Part 11 compliant environment with Thermo Scientific™ LCQUAN™ quantitative software.
Have Questions?
Catalog number LCQUAN25
Price (USD)/ Each
-
Request A Quote

Acquire and manage quantitative LC-MS data in a 21 CFR Part 11 compliant environment using Thermo Scientific™ LCQUAN™ quantitative software. Designed with extensive customer input, LCQUAN quantitative software is a complete, secure data system that allows regulated laboratories to submit documents, including those with electronic signatures, to the FDA in electronic form.

Productive
  • Provides a step-by-step workflow to simplify data acquisition and review
  • Performs method development, data review, processing, reporting, and data export
  • Imports sequence information from other systems
Secure Data Storage
  • Safely processes data stored on networked computes, including secure remote disks
  • Ensures easy data archival and retrieval of relevant project files in a workbook-style, centralized file location
  • Provides for multi-level security with configurable user privileges ranging from full system to data review only access
  • Supports bioanalytical studies in drug development with a secure, digital, bi-directional interface with Thermo Scientific™ Watson LIMS™ laboratory information management system
Specifications, terms, and pricing are subject to change. Not all products are available in all countries. Please consult your local sales representative for details.
Specifications
TypeSoftware
Certifications/Compliance21 CFR Part 11
DescriptionLCquan quantitative software, Configurable
For Use With (Equipment)LC-MS Systems
Unit SizeEach