Customize refrigerator and freezer configurations to better suit the needs of the lab with Thermo Scientific™ Refrigerator and Freezer Door Options which include glass and heated glass doors, as well as either right- or left-hand swing.
Catalog Number
LE7013
Unit Size
Each
Type
Refrigerator and Freezer Door, Glass
Hinge Side
Right
For Use With (Equipment)
29.2 cu. ft. (826L) Forma, Jewett, Puffer Hubbard, and Revco High Performance
11.5 cu. ft. (326L) Forma, Jewett, Puffer Hubbard, and Revco High Performance
Unit Size
Each
Showing 10 of 26Show more
For use with Thermo Scientific™ Forma™, Jewett™, Puffer Hubbard™ and Revco™ series. Solid doors are standard on most refrigerators unless otherwise specified. Optional dual-pane glass doors must be specified when ordering. Contact your Sales Representative for information about heated glass doors.
Glass doors standard on selected blood bank, chromatography and pharmacy refrigerators
Sliding glass doors standard on all 45.8 cu. ft. (1297L) models
Heated glass doors increase amperage and may require next size breaker
Ordering Information:
Door options must be specified at time of order
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.