MAS™ Liquimmune™ Immunoassay Controls
MAS™ Liquimmune™ Immunoassay Controls
Actual product may vary
Thermo Scientific™
MAS™ Liquimmune™ Immunoassay Controls
Get relief from time consuming and costly QC standard lot changes for up to 5 years with ready-to-use, liquid assayed Thermo Scientific™ MAS™ Liquimmune™ Immunoassay Controls.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionLevel
LIG-303Liquimmune Controls, Assayed3
LIG-202Liquimmune Controls, Assayed2
10013876Liquimmune Controls, Vista Vials, Assayed1
10013877Liquimmune Controls, Vista Vials, Assayed2
10013878Liquimmune Controls, Vista Vials, Assayed3
LIG-101Liquimmune Controls, Assayed1
Catalog number LIG-303
Price (USD)/ Each
-
Request A Quote
Description:
Liquimmune Controls, Assayed
Level:
3
Get relief from time-consuming and costly QC standard lot changes for up to 5 years with ready-to-use, liquid assayed Thermo Scientific™ MAS™ Liquimmune™ Immunoassay Controls. With a new 5-year shelf life, these multi-analyte controls simplify QC while monitoring immunoassay test procedures on most automated instruments. These controls are also available in a convenient Vista™ vial format. Extended claim may not apply for all countries.
  • Features three significant levels
  • Human serum matrix
  • Assayed values available for most automated instruments
  • Extended 5-year shelf life beginning with lot LIA2004 in Fall 2015
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionLiquimmune Controls, Assayed
Detectable Analytes17-a-hydroxyprogesterone, Acetaminophen, alpha-Fetoprotein (AFP), Aldosterone, Amikacin, Benzodiazepines, Carbamazepine, Carcinoembryonic Antigen (CEA), Creatinine Kinase-MB (CK-MB), Cortisol, DHEA Sulfate, Digoxin, Disopyramide, Estradiol, Ethosuximide, Ferritin, Folate (Vitamin B9), Follicle Stimulating Hormone (FSH), Gentamicin, Growth Hormone, β-hCG, hCG, Homocysteine, IgA, IgE, IgM, LD-1, Luteinizing Hormone (LH), Lidocaine, Lithium, N-acetylprocainamide (NAPA), Phenobarbital, Phenytoin, Primidone, Procainamide, Progesterone, Prolactin, PSA, PSA (Free), Quinidine, Salicylate (SALY), T-uptake, T3, T3 (Free), T4, Testosterone, Theophylline, Thyroglobulin, Tobramycin, Tricyclic Antidepressants, Thyroid Stimulating Hormone (TSH), Valproic Acid, Vancomycin, Vitamin B12
Sample TypeSerum
TypeControl
Level3
Packaging6 Pk.
Quantity5 mL
Unit SizeEach
Contents & Storage
3-year shelf life from date of manufacture at -25°C to -15°C; 90 day unopened-vial stability for all analytes at 2°C to 8°C; 30 day open-vial stability for all analytes at 2°C to 8°C. Extended 5-year shelf life beginning with lot LIA2004 in Fall 2015.