Invitrogen™

Lipofectamine™ MessengerMAX™ Transfection Reagent

Catalog number: LMRNA150
Have Questions?
Invitrogen™

Lipofectamine™ MessengerMAX™ Transfection Reagent

Catalog number: LMRNA150
Have Questions?
Catalog Number
LMRNA150
Unit Size
15 ml
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
0.1 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
0.3 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
0.75 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1.5 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
LMRNA15015 ml
-
LMRNA0010.1 mL
-
LMRNA0030.3 mL
-
LMRNA0080.75 mL
-
LMRNA0151.5 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Lipofectamine™ MessengerMAX mRNA Transfection Reagent delivers amazing transfection efficiency in neurons and a broad-spectrum of primary cells, enabling improved application outcomes and more biologically relevant research. That's because our novel lipid nanoparticle technology is optimized to deliver the highest amount of mRNA possible without the nuclear entry step that is required with DNA.

Successfully transfect more predictive cell models with a reagent that provides:
• Amazing transfection efficiency in neurons and primary cell types
• Faster protein expression with no risk of genomic integration
• Up to 10x higher cleavage using mRNA CRISPRs

Enable high transfection efficiency in novel genome editing applications
Lipofectamine™ MessengerMAX™ reagent helps increase the likelihood of successful cleavage and recombination with GeneArt™ CRISPR Nuclease mRNA through highly efficient transfection, ultimately maximizing the efficiency of genetic modifications and simplifying the downstream processes.

Efficiently transfect difficult-to-transfect cells
Lipofectamine™ MessengerMAX™ reagent is designed to transfect a higher amount of mRNA into neurons and a broad spectrum of difficult-to-transfect primary cells. This eliminates the need for electroporation or viruses, and provides a > 2-fold improvement in transfection efficiency compared to other lipid based reagents.

Leverage advantages of mRNA transfection
Transfection of mRNA does not require nuclear uptake, which results in faster protein expression than DNA transfection since translation of mRNA occurs in the cytoplasm. Also, mRNA has no risk of genomic integration and transfection efficiency is cell cycle independent.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type
Established Cell Lines, Neuronal Cells, Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample Type
mRNA
Transfection Technique
Lipid-based Transfection
For Use With (Application)
Transfection
Green Features
Sustainable packaging
Product Line
Lipofectamine™
Product Type
Transfection Reagent
Quantity
15 mL
Serum Compatible
Yes
Shipping Condition
Room Temperature

Contents & Storage

Store in refrigerator (2–8°C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text