Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Quantity |
---|---|
LP0011T | 5 kg |
LP0011B | 500 g |
LP0011K | 25 kg |
Oxoid Agar Bacteriological (Agar No. 1) is a clear purified high gel strength bacteriological agar compatible with all culture media.
Share catalog number, name or link