Oxoid™ Yeast Extract Powder
Oxoid™ Yeast Extract Powder
Actual product may vary
Thermo Scientific™
Oxoid™ Yeast Extract Powder
Oxoid Yeast Extract Powder is a dried yeast autolysate which provides a nutritious source of nitrogen and vitamins.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
LP0021T5 kg
LP0021B500 g
2 Options
Catalog number LP0021T
Price (USD)/ Each
-
Request A Quote
Quantity:
5 kg
Oxoid Yeast Extract Powder is a dried yeast autolysate which provides a nutritious source of nitrogen and vitamins.
  • Recommended for plate count of organisms in water
  • Increases yield of organisms of many media or fermentation broths
Specifications
DescriptionYeast Extract Powder
FormPowder
Product TypeMicrobiology Media
Quantity5 kg
Unit SizeEach