Oxoid™ Bile Salts
Oxoid™ Bile Salts
Thermo Scientific™
Oxoid™ Bile Salts
Oxoid Bile Salts is a standardized ox-bile extract used as a selective inhibitory agent in culture media.
Have Questions?
變更視圖buttonViewtableView
產品號碼Quantity
LP0055J250 g
LP0055T5 kg
產品號碼 LP0055J
價格 (HKD)
-
申請報價
Quantity:
250 g
Ask our AI about this Product
Oxoid Bile Salts is a standardized ox-bile extract used as a selective inhibitory agent in culture media.
Normal working concentration 0.5% w/v. Each vial supplements 500 mL of medium.
規格
DescriptionBile Salts
Quantity250 g
FormPowder
Product TypeBile Salts
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
文件與下載
證書
批號Certificate TypeDateCatalog Number(s)
4291390-02Certificate of AnalysisJul 02, 2025LP0055J
4502666Certificate of AnalysisJun 14, 2025LP0055J
4480888-02Certificate of AnalysisJun 14, 2025LP0055J
4480888Certificate of AnalysisJun 14, 2025LP0055J
4446731-02Certificate of AnalysisDec 10, 2024LP0055J
顯示 5 個結果, 在上方搜尋特定憑證
安全資料表
分享目錄編號、產品名稱或連結。
1x1 image pixel for data collection