Oxoid™ Bile Salts No. 3
Oxoid™ Bile Salts No. 3
Thermo Scientific™
Oxoid™ Bile Salts No. 3
The Oxoid Bile Salts No. 3 is a specifically refined fraction of bile acid salts.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
LP0056J250 g
LP0056L20 kg
Catalog number LP0056J
Price (USD)
-
Request A Quote
Quantity:
250 g
Ask our AI about this Product
Oxoid Bile Salts No. 3 is a specifically refined fraction of bile acid salts.
Normal working concentration 0.15% w/v. Each vial supplements 500 mL of medium.
Specifications
DescriptionBile Salts No. 3
Quantity250 g
FormPowder
Product TypeBile Salts
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Lot #Certificate TypeDateCatalog Number(s)
6271363Certificate of AnalysisJun 14, 2025LP0056J
6246558Certificate of AnalysisApr 10, 2025LP0056J
6186125Certificate of AnalysisNov 27, 2024LP0056J
6132044Certificate of AnalysisJul 23, 2024LP0056J
3669177Certificate of AnalysisOct 20, 2023LP0056J
5 results displayed, search above for a specific certificate
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection