Premixed Mobile Phase Solutions
Premixed liquid solutions of various chemical compositions formulated for use as mobile phases in chromatography and mass spectrometry applications. The mobile phase dissolves chromatographic samples and carries them through a stationary phase for separation.