Search Thermo Fisher Scientific
Connector Types | Includes wall plugs for US/Japan, Euro, UK/Singapore, Australia/New Zealand and China wall outlets |
Description | Orion Lab Star series 100-240 V 50/60 Hz universal power adapter with twist-and-lock connector |
Type | Accessory |
Certifications/Compliance | CE, CSA, FCC Class A limits |
Compatibility | Orion Lab Star PH111 pH meter, Orion Lab Star EC112 conductivity meter and Orion Lab Star DO113 DO meter |
Unit Size | Each |
Description | Orion Lab Star series electrode stand with meter-attachable bracket |
Type | Accessory |
Certifications/Compliance | None |
Compatibility | Orion Lab Star PH111 pH meter, Orion Lab Star EC112 conductivity meter and Orion Lab Star DO113 DO meter |
Unit Size | Each |
Description | Orion Lab Star series meter dust cover |
Type | Accessory |
Certifications/Compliance | None |
Compatibility | Orion Lab Star PH111 pH meter, Orion Lab Star EC112 conductivity meter and Orion Lab Star DO113 DO meter |
Unit Size | Each |
Description | Orion Lab Star series USB computer cable |
Type | Accessory |
Certifications/Compliance | None |
Compatibility | Orion Lab Star PH111 pH meter, Orion Lab Star EC112 conductivity meter and Orion Lab Star DO113 DO meter |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Certifications/Compliance | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
LSTAR-PWR | Each | Orion Lab Star series 100-240 V 50/60 Hz universal power adapter with twist-and-lock connector | CE, CSA, FCC Class A limits | 47.07 Online price 52.30 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
LSTAR-ARM | Each | Orion Lab Star series electrode stand with meter-attachable bracket | None | 94.50 Online price 105.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
LSTAR-CVR | Each | Orion Lab Star series meter dust cover | None | 47.07 Online price 52.30 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
LSTAR-USB | Each | Orion Lab Star series USB computer cable | None | 47.07 Online price 52.30 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Orion Lab Star Series Meter Accessories Features
Additional Accessories Compatible with Orion Lab Star Series Meters