Gibco™

Medium 154

Catalog number: M154500
Have Questions?
Gibco™

Medium 154

Catalog number: M154500
Have Questions?
Catalog Number
M154500
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
M154500500 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
A sterile, liquid medium prepared with 200 μM calcium chloride for the culture of human epidermal keratinocytes. This basal medium requires the addition of HKGS (S-001-5) or HKGS Kit (S-001-K) prior to use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type
Keratinocyte Cells
Classification
Serum-free
Culture Type
Mammalian Cell Culture
Form
Liquid
Product Type
Medium 154
Serum Level
Serum-free
Sterility
Sterile-filtered
Sterilization Method
Sterile-filtered
With Additives
Phenol Red
Green Features
Sustainable packaging
Inorganic Salts
Calcium
Product Line
Cascade Biologics™
Shipping Condition
Room Temperature

Contents & Storage

Store at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text