Maxima™ H Minus cDNA Synthesis Master Mix
Maxima™ H Minus cDNA Synthesis Master Mix
Thermo Scientific™

Maxima™ H Minus cDNA Synthesis Master Mix

Thermo Scientific Maxima H Minus cDNA Synthesis Master Mix, available with or without dsDNase, provides all cDNA synthesis reaction components in a convenient one-tube master mix.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludesNo. of Reactions
M1682Kit with dsDNase200 Reactions
M1661Kit only50 Reactions
M1662Kit only200 Reactions
M1681Kit with dsDNase50 Reactions
Catalog number M1682
Price (EUR)
1.704,00
Each
-
Add to cart
Includes:
Kit with dsDNase
No. of Reactions:
200 Reactions
Request bulk or custom format
Price (EUR)
1.704,00
Each
Add to cart
Ask our AI about this Product
Thermo Scientific Maxima H Minus cDNA Synthesis Master Mix, available with or without dsDNase, provides all cDNA synthesis reaction components in a convenient one-tube master mix. It is optimized for highly efficient cDNA synthesis for two-step quantitative RT-PCR (RT-qPCR) applications and reproducible cDNA synthesis at elevated temperatures (50–65°C) within 30 minutes.

The master mix contains Maxima H Minus Reverse Transcriptase (RT) and RiboLock RNase Inhibitor. Maxima H Minus RT is an advanced enzyme derived from M-MuLV RT by in vitro evolution. The enzyme features high thermostability, robustness, processivity, increased cDNA synthesis rate, and lack of RNase H activity. The recombinant RiboLock RNase Inhibitor effectively protects RNA template from degradation by RNases A, B, and C at temperatures up to 55°C. The master mix also contains reaction buffer, dNTPs, oligo (dT)18, and random hexamer primers. A separate tube of No-RT control mix is also provided for convienient RT negative control.

Both kits are capable of reproducible cDNA synthesis at elevated temperatures (50°C–65°C). The synthesis reaction is typically complete in 15–30 minutes. The included double-strand specific DNase in the Maxima H Minus cDNA Synthesis Master Mix with dsDNase allows removal of genomic DNA from RNA samples in two minutes without affecting the quality or quantity of RNA. This dsDNase is also available separately (Cat. No. EN0771).

Features of Maxima H Minus cDNA Synthesis Master Mix include:
• One-tube master mix helps reduce pipetting steps and enhanced consistency in RT-qPCR results
• Increased RT efficiency across a wide range of input RNA amounts and gene targets
• High thermostability to allow RT reactions at 50–65°C temperature range
• Integrated step of genomic DNA removal from RNA samples (Maxima H Minus cDNA Synthesis Master Mix with dsDNase only)

Additional information about reaction components
• The No RT control mix contains all the components in the Maxima H Minus cDNA Synthesis Master Mix except RT. The presence of a PCR product in the No RT control reaction indicates that the reaction is contaminated with DNA. To further enhance genomic DNA elimination efficiency, template RNA incubation with dsDNase (Cat. No. EN0771) is strongly recommended.
• Nuclease-free water is provided for reaction setup and dilution of sample DNA. The absence of exodeoxyribonucleases, ribonucleases, and phosphatases has been confirmed by appropriate quality tests.
• To further enhance genomic DNA elimination efficiency, template RNA incubation with the included dsDNase is strongly recommended. (Maxima H Minus cDNA Synthesis Master Mix with dsDNase only)
• The dsDNase is used for rapid and safe removal of contaminating genomic DNA from RNA samples. dsDNase is easily inactivated by moderate heat treatment (55°C). It allows for a dramatically simplified workflow that combines genomic DNA elimination and cDNA synthesis into one-tube procedure. (Maxima H Minus cDNA Synthesis Master Mix with dsDNase only)

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeFirst-Strand cDNA
FormatMaster Mix
IncludesKit with dsDNase
No. of Reactions200 Reactions
Optimal Reaction Temperature50°C
QuantityEach
Reaction FormatPremixed Components
Reagent TypeReverse Transcription
Reverse TranscriptaseMaxima H Minus
Ribonuclease H ActivityReduced
Shipping ConditionDry Ice
Size (Final Product)Up to 20 kb
Starting MaterialRNA
TechniqueReverse Transcription
For Use With (Application)Real Time PCR (qPCR), RT-PCR
GC-Rich PCR PerformanceHigh
Reaction Speed30 min.
Unit SizeEach
Contents & Storage

• Maxima H Minus cDNA Synthesis Master Mix (1 x 800 μL)
• Maxima H Minus cDNA Synthesis Master Mix 'No RT' Control (1 x 200 μL)
• dsDNase (1 x 200 μL)
• 10X dsDNase buffer (1 x 400 μL)
• Nuclease-free water (1 x 1.25 mL)

Store at –5 to –30°C.

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link