Medium 254
Medium 254
Gibco™

Medium 254

Medium 254 is a sterile, liquid tissue culture medium prepared with 200 μM calcium chloride for the culture of normalRead more
Have Questions?
Catalog NumberQuantity
M254500500 mL
Catalog number M254500
Price (KRW)
142,000
Each
-
Add to cart
Quantity:
500 mL
Price (KRW)
142,000
Each
Add to cart
Ask our AI about this Product
Medium 254 is a sterile, liquid tissue culture medium prepared with 200 μM calcium chloride for the culture of normal human epidermal melanocytes. This medium is intended for use as one component in a complete culture environment. When supplemented with HMGS or HMGS-2, Medium 254 will support the plating and proliferation of melanocytes at varying culture densities from 5 x 103 cells/cm2 to 1 x 105 cells/cm2. Additional applications for use may include primary isolation of melanocytes from skin.

Medium 254 is a basal medium containing essential and non-essential amino acids, vitamins, other organic compounds, trace minerals, and inorganic salts. This medium does not contain antibiotics, antimycotics, hormones, growth factors, or proteins. This medium is HEPES and bicarbonate buffered and designed for use in an incubator with an atmosphere of 5% CO2/95% air. To support plating and long-term proliferation of normal human melanocytes, Medium 254 must be supplemented with Human Melanocyte Growth Supplement (HMGS) or PMA-Free Human Melanocyte Growth Supplement-2 (HMGS-2). Each of these supplements contains all of the growth factors, hormones, and tissue extracts necessary for growth of melanocytes in Medium 254.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeMelanocytes
Inorganic SaltsCalcium
Product LineCascade Biologics™
Product TypeMedium 254
Quantity500 mL
Shipping ConditionRoom Temperature
Culture TypeMammalian Cell Culture
FormLiquid
SterilitySterile-filtered
Sterilization MethodSterile-filtered
With AdditivesHEPES, Phenol Red
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.
Media Formulations
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link