MitoTracker™ Dyes for Mitochondria Labeling
MitoTracker™ Dyes for Mitochondria Labeling
Invitrogen™

MitoTracker™ Dyes for Mitochondria Labeling

These mitochondria dyes are specially packaged in vials of 50 μg each to create freshly reconstituted dyes without the impact of freeze-thaw cycles to aid in a simplified sample prep for cell analysis workflow.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionExcitation Wavelength Range
M7511MitoTracker Orange CM-H2TMRos554/576 nm
M22426MitoTracker Deep Red FM644/665 nm
M22425MitoTracker™ Red FM581/644 nm
M7512MitoTracker™ Red CMXRos579/599 nm
M7513MitoTracker Red CM-H2Xros579/599 nm
M7514MitoTracker™ Green FM490/516 nm
M7510MitoTracker Orange CMTMRos554/576 nm
Catalog number M7511
Price (USD)
437.88
Special offer
Online exclusive
Ends: 27-Mar-2026
534.00
Save 96.12 (18%)
Each
Add to cart
Description:
MitoTracker Orange CM-H2TMRos
Excitation Wavelength Range:
554/576 nm
Price (USD)
437.88
Special offer
Online exclusive
Ends: 27-Mar-2026
534.00
Save 96.12 (18%)
Each
Add to cart

Rosamine & carbocyanine-based staining dyes MitoTracker Orange CMTMRos, Red CMXRos, Orange CM-H2TMRos, Red CM-H2XRos, Red FM, Green FM, & Deep Red FM enable mitochondria visualization with fluorescent imaging. These mitochondria dyes are specially packaged in vials of 50 μg each to create freshly reconstituted dyes without the impact of freeze-thaw cycles to aid in a simplified sample prep for cell analysis workflow.

Rosamine-based MitoTracker dyes for mitochondria labeling

Cell-permeant MitoTracker probes stain active mitochondria in live cells for labeling and localization in fluorescent cell imaging. Our rosamine-based mitochondrial staining dyes include MitoTracker Orange CMTMRos, a derivative of tetramethylrosamine, and MitoTracker Red CMXRos, a derivative of X-rosamine. MitoTracker Orange CM-H2TMRos and MitoTracker Red CM-H2XRos are reduced, nonfluorescent versions of the rosamine-based MitoTracker dyes that fluoresce upon oxidation. The fluorescent signal from the rosamine-based MitoTracker dyes is retained in the mitochondria even after aldehyde fixation and detergent permeabilization, making these mitochondria dyes flexible for many workflows including applications that require subsequent processing such as immunocytochemistry or in situ hybridization. MitoTrackerRed and Orange are well suited for multicolor labeling experiments because their fluorescence is well resolved from the green fluorescence of other probes.

Carbocyanine-based MitoTracker dyes for mitochondria labeling

Cell-permeant MitoTracker dyes stain active mitochondria in live cells for mitochondrial labeling and localization in fluorescent cell imaging. The carbocyanine-based dyes MitoTracker Green FM and MitoTracker Red FM accumulate in active mitochondria but are not well-retained in mitochondria after aldehyde fixation. The carbocyanine-based MitoTracker Deep Red FM stains active mitochondrial in live cells and is well-retained in mitochondria after aldehyde fixation and subsequent permeabilization with detergents for applications that require subsequent processing such as immunocytochemistry or in situ hybridization. MitoTracker Red FM and MitoTracker Deep Red FM are well suited for multicolor labeling experiments because their red fluorescence is well resolved from the green fluorescence of other probes.

Benefits of using MitoTracker mitochondria labeling dyes

MitoTracker dyes are provided in vials of 50 μg of lyophilized powder ready for reconstitution. To label mitochondria, live cells are simply incubated with the MitoTracker probe of your choice. The mitochondrial staining dyes passively diffuse across the plasma membrane and accumulate in active mitochondria. The MitoTracker dyes are offered in a range of wavelengths and can be used for mitochondrial localization in multicolor experiments.

Conventional fluorescent stains such as tetramethylrosamine and rhodamine 123 are readily sequestered by active mitochondria and are reversible in dynamic membrane potential measurements as they easily wash out of cells upon loss in membrane potential. In contrast, MitoTracker dyes contain a mildly thiol-reactive chloromethyl moiety so that mitochondrial staining is retained if the mitochondrial membrane potential is lost, allowing many of the MitoTracker dyes to be retained during cell fixation.

Throughout the cell life cycle, mitochondria use oxidizable substrates to produce an electrochemical proton gradient across the mitochondrial membrane (whose potential is negative), resulting in ATP production. MitoTracker dyes are ideal probes for mitochondria staining in experiments studying the cell cycle or processes such as apoptosis and other end point assays. MitoTracker dyes are also available for flow cytometry applications (Cat. No. M46750, M46751, M46752, and M46753).

Related products for mitochondrial membrane potential

For studying dynamic mitochondria membrane potential specifically, we recommend using JC-1 (cationic carbocyanine dye, Cat. No. T3168) or TMRM (tetramethyl rhodamine methyl ester, Cat. No. I34361) dyes. The MitoProbe JC-1 Assay Kit (Cat. No. M34152) contains the JC-1 dye in addition to the potent mitochondrial membrane-potential disrupter, CCCP, which depolarizes the mitochondrial membrane. These reagents can provide compensatory controls to correctly compensate green-to-red fluorescence ratio. TMRM is used for the detection of dynamic measurement of mitochondrial membrane potential.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorOrange
DescriptionMitoTracker Orange CM-H2TMRos
Detection MethodFluorescence
Excitation Wavelength Range554/576 nm
For Use With (Equipment)Fluorescence Microscope, Flow Cytometer, Microplate Reader
FormatSpecial packaging
Product LineMitoTracker
Quantity20 x 50 μg
Shipping ConditionRoom Temperature
Label TypeFluorescent Dye
Product TypeDye
SubCellular LocalizationMitochondria/Organelle Stain
Unit SizeEach
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.

Frequently asked questions (FAQs)

It looks like my Mitotracker dye is staining more than just the mitochondria. Why?

This is typically a result of using too high of a concentration of the Mitotracker dye. Most organic dyes are used in the low micromolar range. The MitoTracker dyes are used at a much lower concentration, around 50–200 nanomolar. Higher concentrations can cause background fluorescence and non-mitochondrial staining.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I am testing mitochondrial membrane potential, but my untreated cells are fluorescing, and I'm not seeing a significant difference in my test sample.

Regardless of which dye you use - tetramethylrhodamine, methyl ester (TMRM), JC-1 or MitoTracker - untreated cells will fluoresce. It's just that cells with reduced mitochondrial membrane potential will fluoresce less. It is the degree of change which is important. JC-1 dye not only changes intensity, but has a ratiometric spectral change in excitation and emission. It is very important to have an untreated control as well as a positive control treated with a mitochondrial membrane potential destabilizer, such as CCCP or FCCP. Most mitochondrial stains are only for use with live cells, as the signal will not be retained to the same degree with fixation.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (33)

Citations & References
Abstract
Bcl-2 controls caspase activation following a p53-dependent cyclin D1-induced death signal.
Authors:Pratt MA, Niu MY
Journal:J Biol Chem
PubMed ID:12480939
'MCF-7 and ZR-75 breast cancer cells infected with an adenovirus constitutively expressing high levels of cyclin D1 demonstrated widespread mitochondrial translocation of Bax and cytochrome c release that was approximately doubled after the addition of all-trans retinoic acid (RA) or Bcl-2 antisense oligonucleotide. By comparison, the percentage of cells in ... More
Ketamine induces toxicity in human neurons differentiated from embryonic stem cells via mitochondrial apoptosis pathway.
Authors:Bosnjak ZJ, Yan Y, Canfield S, Muravyeva MY, Kikuchi C, Wells CW, Corbett JA, Bai X,
Journal:Curr Drug Saf
PubMed ID:22873495
'Ketamine is widely used for anesthesia in pediatric patients. Growing evidence indicates that ketamine causes neurotoxicity in a variety of developing animal models. Our understanding of anesthesia neurotoxicity in humans is currently limited by difficulties in obtaining neurons and performing developmental toxicity studies in fetal and pediatric populations. It may ... More
Spatiotemporal compartmentalization of key physiological processes during muscle precursor differentiation.
Authors:Ozbudak EM, Tassy O, Pourquié O,
Journal:Proc Natl Acad Sci U S A
PubMed ID:20160088
'The development of multicellular organisms is controlled by transcriptional networks. Understanding the role of these networks requires a full understanding of transcriptome regulation during embryogenesis. Several microarray studies have characterized the temporal evolution of the transcriptome during development in different organisms [Wang QT, et al. (2004) Dev Cell 6:133-144; Furlong ... More
Mutant huntingtin expression induces mitochondrial calcium handling defects in clonal striatal cells: functional consequences.
Authors:Milakovic T, Quintanilla RA, Johnson GV
Journal:J Biol Chem
PubMed ID:16973623
'Huntington disease (HD) is caused by a pathological elongation of CAG repeats in the huntingtin protein gene and is characterized by atrophy and neuronal loss primarily in the striatum. Mitochondrial dysfunction and impaired Ca2+ homeostasis in HD have been suggested previously. Here, we elucidate the effects of Ca2+ on mitochondria ... More
Motor deficit in a Drosophila model of mucolipidosis type IV due to defective clearance of apoptotic cells.
Authors:Venkatachalam K, Long AA, Elsaesser R, Nikolaeva D, Broadie K, Montell C,
Journal:Cell
PubMed ID:19041749
'Disruption of the Transient Receptor Potential (TRP) mucolipin 1 (TRPML1) channel results in the neurodegenerative disorder mucolipidosis type IV (MLIV), a lysosomal storage disease with severe motor impairments. The mechanisms underlying MLIV are poorly understood and there is no treatment. Here, we report a Drosophila MLIV model, which recapitulates the ... More