MitoTracker™ Dyes for Mitochondria Labeling
MitoTracker™ Dyes for Mitochondria Labeling
Invitrogen™

MitoTracker™ Dyes for Mitochondria Labeling

These mitochondria dyes are specially packaged in vials of 50 μg each to create freshly reconstituted dyes without the impact of freeze-thaw cycles to aid in a simplified sample prep for cell analysis workflow.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionExcitation Wavelength Range
M7512MitoTracker™ Red CMXRos579/599 nm
M22426MitoTracker Deep Red FM644/665 nm
M22425MitoTracker™ Red FM581/644 nm
M7513MitoTracker Red CM-H2Xros579/599 nm
M7514MitoTracker™ Green FM490/516 nm
M7510MitoTracker Orange CMTMRos554/576 nm
M7511MitoTracker Orange CM-H2TMRos554/576 nm
Catalog number M7512
Price (USD)
444.44
Special offer
Online exclusive
Ends: 27-Mar-2026
542.00
Save 97.56 (18%)
Each
Add to cart
Description:
MitoTracker™ Red CMXRos
Excitation Wavelength Range:
579/599 nm
Price (USD)
444.44
Special offer
Online exclusive
Ends: 27-Mar-2026
542.00
Save 97.56 (18%)
Each
Add to cart

Rosamine & carbocyanine-based staining dyes MitoTracker Orange CMTMRos, Red CMXRos, Orange CM-H2TMRos, Red CM-H2XRos, Red FM, Green FM, & Deep Red FM enable mitochondria visualization with fluorescent imaging. These mitochondria dyes are specially packaged in vials of 50 μg each to create freshly reconstituted dyes without the impact of freeze-thaw cycles to aid in a simplified sample prep for cell analysis workflow.

Rosamine-based MitoTracker dyes for mitochondria labeling

Cell-permeant MitoTracker probes stain active mitochondria in live cells for labeling and localization in fluorescent cell imaging. Our rosamine-based mitochondrial staining dyes include MitoTracker Orange CMTMRos, a derivative of tetramethylrosamine, and MitoTracker Red CMXRos, a derivative of X-rosamine. MitoTracker Orange CM-H2TMRos and MitoTracker Red CM-H2XRos are reduced, nonfluorescent versions of the rosamine-based MitoTracker dyes that fluoresce upon oxidation. The fluorescent signal from the rosamine-based MitoTracker dyes is retained in the mitochondria even after aldehyde fixation and detergent permeabilization, making these mitochondria dyes flexible for many workflows including applications that require subsequent processing such as immunocytochemistry or in situ hybridization. MitoTrackerRed and Orange are well suited for multicolor labeling experiments because their fluorescence is well resolved from the green fluorescence of other probes.

Carbocyanine-based MitoTracker dyes for mitochondria labeling

Cell-permeant MitoTracker dyes stain active mitochondria in live cells for mitochondrial labeling and localization in fluorescent cell imaging. The carbocyanine-based dyes MitoTracker Green FM and MitoTracker Red FM accumulate in active mitochondria but are not well-retained in mitochondria after aldehyde fixation. The carbocyanine-based MitoTracker Deep Red FM stains active mitochondrial in live cells and is well-retained in mitochondria after aldehyde fixation and subsequent permeabilization with detergents for applications that require subsequent processing such as immunocytochemistry or in situ hybridization. MitoTracker Red FM and MitoTracker Deep Red FM are well suited for multicolor labeling experiments because their red fluorescence is well resolved from the green fluorescence of other probes.

Benefits of using MitoTracker mitochondria labeling dyes

MitoTracker dyes are provided in vials of 50 μg of lyophilized powder ready for reconstitution. To label mitochondria, live cells are simply incubated with the MitoTracker probe of your choice. The mitochondrial staining dyes passively diffuse across the plasma membrane and accumulate in active mitochondria. The MitoTracker dyes are offered in a range of wavelengths and can be used for mitochondrial localization in multicolor experiments.

Conventional fluorescent stains such as tetramethylrosamine and rhodamine 123 are readily sequestered by active mitochondria and are reversible in dynamic membrane potential measurements as they easily wash out of cells upon loss in membrane potential. In contrast, MitoTracker dyes contain a mildly thiol-reactive chloromethyl moiety so that mitochondrial staining is retained if the mitochondrial membrane potential is lost, allowing many of the MitoTracker dyes to be retained during cell fixation.

Throughout the cell life cycle, mitochondria use oxidizable substrates to produce an electrochemical proton gradient across the mitochondrial membrane (whose potential is negative), resulting in ATP production. MitoTracker dyes are ideal probes for mitochondria staining in experiments studying the cell cycle or processes such as apoptosis and other end point assays. MitoTracker dyes are also available for flow cytometry applications (Cat. No. M46750, M46751, M46752, and M46753).

Related products for mitochondrial membrane potential

For studying dynamic mitochondria membrane potential specifically, we recommend using JC-1 (cationic carbocyanine dye, Cat. No. T3168) or TMRM (tetramethyl rhodamine methyl ester, Cat. No. I34361) dyes. The MitoProbe JC-1 Assay Kit (Cat. No. M34152) contains the JC-1 dye in addition to the potent mitochondrial membrane-potential disrupter, CCCP, which depolarizes the mitochondrial membrane. These reagents can provide compensatory controls to correctly compensate green-to-red fluorescence ratio. TMRM is used for the detection of dynamic measurement of mitochondrial membrane potential.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorRed
DescriptionMitoTracker™ Red CMXRos
Detection MethodFluorescence
Excitation Wavelength Range579/599 nm
For Use With (Equipment)Fluorescence Microscope, Flow Cytometer, Microplate Reader
FormatSpecial packaging
Product LineMitoTracker
Quantity20 x 50 μg
Shipping ConditionRoom Temperature
Label TypeFluorescent Dye
Product TypeDye
SubCellular LocalizationMitochondria/Organelle Stain
Unit SizeEach
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.

Frequently asked questions (FAQs)

It looks like my Mitotracker dye is staining more than just the mitochondria. Why?

This is typically a result of using too high of a concentration of the Mitotracker dye. Most organic dyes are used in the low micromolar range. The MitoTracker dyes are used at a much lower concentration, around 50–200 nanomolar. Higher concentrations can cause background fluorescence and non-mitochondrial staining.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can I use MitoTracker Red CMXRos dye in a plate reader to study mitochondrial membrane potential changes?

Yes, it has been done. A literature search will find numerous examples. However, be aware that plate readers are typically less sensitive than microscopes or flow cytometers, which means that the degree of change in fluorescence may not be as sensitive or easily detectable with a plate reader. Be sure to optimize label times and concentrations carefully.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Our facility does not allow flow cytometry of live cells. Can I fix the cells after staining with MitoTracker Red CMXRos dye?

Yes, this is possible because MitoTracker Red CMXRox dye is retained upon fixation. See Table 2 in this manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/mp07510.pdf).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

After I labeled neurons with MitoTracker Red CMXRos, they are dead the next day. Is this expected?

The Mitotracker dyes should be imaged soon after staining because over time, those dyes can be toxic.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What cellular processes can be analyzed with a flow cytometer?

-Calcium flux: Each of the Oregon Green calcium indicators binds intracellular calcium with increasing affinity, providing a sensitivity range to match many applications. Oregon Green probes emit green fluorescence at resting levels of Ca2+ and increase their fluorescence intensity 14-fold with increasing Ca2+ concentration. The cell-permeant formulation (Cat. No. O6807) can be loaded in cell media and is compatible with flow cytometry.
-Rhodamine-based calcium indicators comprise a range of probes for large or small changes in Ca2+ concentration. They exhibit a 50-fold increase in fluorescence upon calcium binding and offer a range of wavelengths that can be used in conjunction with GFP or green-fluorescent dyes for multiplexing. Rhod-2, AM (Cat. No. R1245MP), in particular, localizes to mitochondria and can be used with flow cytometry.
-Membrane potential: A distinctive feature of the early stages of apoptosis is the disruption of the mitochondria, including changes in membrane and redox potential. We offer a range of products specifically designed to assay mitochondrial membrane potential in live cells by flow cytometry, with minimal disruption of cellular function. The MitoProbe family of mitochondrial stains (Cat. Nos. M34150, M34151, and M34152) provide quick, easy, and reliable flow cytometric detection of the loss of mitochondrial membrane potential that occurs during apoptosis. MitoTracker dyes (Cat. Nos. M7510 and M7512) are membrane potential-dependent probes for staining mitochondria in live cells. The staining pattern of MitoTracker dyes is retained throughout subsequent flow cytometry immunocytochemistry, DNA end labeling, in situ hybridization, or counterstaining steps. The Mitochondrial Permeability Transition Pore Assay (Cat. No. M34153) provides a more direct method of measuring mitochondrial permeability transition pore opening than assays relying on mitochondrial membrane potential alone. The mitochondrial permeability transition pore (MPTP) is a non-specific channel formed by components from the inner and outer mitochondrial membranes, and appears to be involved in the release of mitochondrial components during cell death.
-Phagocytosis: In phagocytosis, cells internalize particulate matter such as microorganisms, and this process is important for immune responses and during the clearance of apoptotic cells. Probes for studying phagocytosis include BioParticles indicators—bacteria and yeast labeled with fluorescent dyes.
-Tracking phagocytosis using a quench/wash-based assay can report on simple uptake, or a pH indicator can be used to monitor stages in the pathway. We have no-wash assays labeled with pHrodo Red or Green (Cat. Nos. A10010, P35361, P35364, P35365, P35366, and P35367) and no-wash assays for whole blood (Cat. Nos. A10025, A10026, P35381, and P35382), all suitable for flow cytometry.
-pH changes: Sensitive pH determinations can be made in a physiological range using either fluorescent intensity or ratiometric measurements. pHrodo dyes (Cat. Nos. P35373 and P35372) provide signal intensity modulation from pH 2 to pH 9 and with a choice of fluorescent wavelengths. Tracking internalization of fluorescent dextran is a routine method for analyzing pH changes in cellular compartments. Dextran conjugates of pHrodo dyes (Cat. Nos. P35368 and P10361) provide the most complete solution by allowing discrimination of vesicles from early endosomes to lysosomes, with no quench or wash required.
-Reactive oxygen species: Cells that are environmentally stressed usually contain greatly increased levels of reactive oxygen species (ROS). CellROX reagents are fluorogenic probes developed for the detection and quantitation of ROS in live cells. These cell-permeant reagents are non-fluorescent or very weakly fluorescent in the reduced state; however, when oxidized, they become brightly fluorescent and remain localized within the cell. We offer CellROX Green (Cat. No. C10492), CellROX Orange (Cat. No. C10493), and CellROX Deep Red (Cat. No. C10491) Assay Kits validated for flow cytometry.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (367)

Citations & References
Abstract
Knockdown of mitochondrial heat shock protein 70 promotes progeria-like phenotypes in caenorhabditis elegans.
Authors:Kimura K,Tanaka N,Nakamura N,Takano S,Ohkuma S
Journal:The Journal of biological chemistry
PubMed ID:17189267
p53 mediates nontranscriptional cell death in dopaminergic cells in response to proteasome inhibition.
Authors:Nair VD,McNaught KS,González-Maeso J,Sealfon SC,Olanow CW
Journal:The Journal of biological chemistry
PubMed ID:17060322
Authors:
Journal:
PubMed ID:9030618
The mitochondrial death/life regulator in apoptosis and necrosis.
Authors:Kroemer G,Dallaporta B,Resche-Rigon M
Journal:Annual review of physiology
PubMed ID:9558479
Both physiological cell death (apoptosis) and, in some cases, accidental cell death (necrosis) involve a two-step process. At a first level, numerous physiological and some pathological stimuli trigger an increase in mitochondrial membrane permeability. The mitochondria release apoptogenic factors through the outer membrane and dissipate the electrochemical gradient of the ... More
Control of mitochondrial structure and function by the Yorkie/YAP oncogenic pathway.
Authors:Nagaraj R, Gururaja-Rao S, Jones KT, Slattery M, Negre N, Braas D, Christofk H, White KP, Mann R, Banerjee U,
Journal:Genes Dev
PubMed ID:22925885
Mitochondrial structure and function are highly dynamic, but the potential roles for cell signaling pathways in influencing these properties are not fully understood. Reduced mitochondrial function has been shown to cause cell cycle arrest, and a direct role of signaling pathways in controlling mitochondrial function during development and disease is ... More