Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: CI2Z9E .
FIGURE: 1 / 4
Immunogen sequence: SCDDCGRSFR LERFLRAHQR QHTGERPFTC AECGKNFGKK THLVAHSRVH SGERPFACEE CGRRFSQGSH LAAHRRDHAP DRPFVCPDCG KAFRHKPYLA AHRRIHTGEK PYVCPDCGKA FSQKSNLVSH RRIHTGERPY ACPDCDRSFS QKSNLITHRK SHIRDGAFCC AICGQTFDDE ERLLAHQKKH DV
Replication initiator 1 (REPIN1) is a gene encoding a zinc-finger protein that plays a critical role in various metabolic processes. Located on chromosome 7 in humans, REPIN1 is prominently expressed in liver and adipose tissues. It is associated with the regulation of body weight and triglyceride levels, as evidenced by its locus within a quantitative trait locus (QTL) related to these metabolic traits. The REPIN1 protein is involved in enhancing DNA bending, which is crucial for replication initiation. Studies have shown that deleting REPIN1 in the liver can lead to improved insulin sensitivity and altered lipid metabolism, with secondary effects on adipose tissue. Additionally, REPIN1 is implicated in iron metabolism and apoptosis regulation, further underscoring its multifaceted role in cellular processes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 60 kDa origin-specific DNA-binding protein; 60 kDa replication initiation region protein; ATT-binding protein; DHFR oribeta-binding protein RIP60; DNA-binding protein REPIN1; H_DJ0584D14.12; replication initiation region protein (60kD); Zinc finger protein 464; zinc finger protein 464 (RIP60); zinc finger protein AP4
Gene Aliases: AP4; REPIN1; RIP60; Zfp464; ZNF464
UniProt ID: (Human) Q9BWE0
Entrez Gene ID: (Human) 29803
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support