Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Enabling real time, accurate results for process monitoring
New cGMP & 21 CFR Part 11 compliant software available.
Depth (Metric) | 25 cm |
Electrical Requirements | Hertz: 50/60 Hz |
Height (Metric) | 7.7 cm |
Laser Power | Laser Excitation Wavelength: 785 nm, Excitation power: 10 to 450 mW |
Operating Temperature Range | 15°C to 33°C |
Resolution | Spectral Resolution: 6.5 cm-1 average across spectral range |
Software Details | MarqMetrix Raman suite Controller: Integrated PC (Windows 10 OS) |
Spectral Range | 100 to 3250 cm-1 |
Width (Metric) | 25 cm |
Safety Features | 1 keyed Laser interlock switch, Electronic laser trigger |
Detector | Thermally stabilized CCD (2 stage TEC) |
Fiber Conditions | FC/PC (Both excitation and collection) |
Includes | Wireless mouse, HDMI cable, monitor |
Certifications/Compliance | Laser Certification: Class 3B laser deviceReported to the FDA |
Dark Noise | ≤16.5 counts at 10 sec. integration |
Ports | 1RJ-45 Ethernet port 3 USB 3.0 |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
MARQMETRIXAIO | Each | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
With the MarqMetrix All-In-One Process Raman Analyzer, businesses can conduct real-time, non-destructive, and immediate analysis without sample preparation.
The MarqMetrix All-In-One Process Raman Analyzer makes precise and accurate measurements simple to execute and understand. Our one-button hardware and easy-to-use software will have you taking highly accurate Raman measurements in less than 15 minutes out of the box.
The MarqMetrix All-In-One Process Raman Analyzer is easily integrated into your existing process and eliminates the need for costly technical expertise.
Application versatility
Other specifications
Use Raman inline for process monitoring
With a small footprint and lower cost, the MarqMetrix All-In-One Process Raman Analyzer makes process monitoring possible in-line and enables you to make optimization decisions faster.
Pack your MarqMetrix All-In-One Process Raman Analyzer in a protective case and take it to the point of need. Factory calibration ensures continuous and precise analysis out of the box.
Probes for multiple applications
Several probes are available for the MarqMetrix All-In-One Process Raman Analyzer. Upon ordering you will choose the probe to best meet your application needs:
Software
Maximize the power of the MarqMetrix All-In-One Process Raman Analyzer with Thermo Scientific Lykos PAT Software – a remote control and monitoring software designed to simplify the complexities of chemometrics with features including:
Service Plans
Thermo Fisher Scientific offers Service Plans designed to maximize productivity and optimize performance throughout the lifecycle of your MarqMetrix All-In-One Process Raman Analyzer. Plans include exclusive service advantages such as:
Speak with your sales representative to learn more about service and support options.