Microbact™ GNB Kit
Microbact™ GNB Kit
Actual product may vary
Microbact™ GNB Kit
Microbact™ GNB Kit
Thermo Scientific™
Microbact™ GNB Kit
Simplify the identification of Enterobacteriaceae and common miscellaneous Gram-negative bacilli with this biochemical identification system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionQuantity
MB1133AMicrobact GNB 12B Kit60 Tests/Kit
MB1131AMicrobact GNB 24E Kit40 Tests/Kit
MB1132AMicrobact GNB 12A Kit60 Tests/Kit
Catalog number MB1133A
Price (USD)
-
Request A Quote
Description:
Microbact GNB 12B Kit
Quantity:
60 Tests/Kit
Ask our AI about this Product
Simplify the identification of aerobic and facultatively anaerobic Gram-negative bacteria (Enterobacteriaceae and miscellaneous Gram-negative bacteria) with the Thermo Scientific™ Oxoid™ Microbact™ GNB Kit – a complete, self-contained biochemical based identification system. The kit uses 24 different biochemical tests in microplate format to produce easy-to read, distinct color reactions, generally following overnight incubation.
Gram-negative bacteria are of significant concern in both clinical infections and food contamination.  Identification of the causative agent is crucial in the clinical setting to identify the cause of infection and to establish an appropriate treatment regime. In the food industry, biochemical identification can help to trace the source of contamination. With this GNB kit, Enterobacteriaceae and common miscellaneous Gram-negative bacilli are quickly and accurately identified so that proper, fast and appropriate action may follow.
  • Fast: Most results available overnight
  • Simple and Economical: Simple microplate format
  • Easy-to-read and Interpret: Results are clearly visible as distinct color reactions that can be interpreted using the Microbact Identification Package (MB1244A)
  • Comprehensive: Identifies 100 Gram-negative bacteria, more unusual Enterobaceriaceae and oxidase-positive, Gram-negative bacteria.
  • Flexible: Use single strips or a combined system for more complex identifications. Ideal for use in clinical or food microbiology laboratories.
  • Reliable: A comprehensive range of biochemical tests based on published reference methodologies1-4
Additional items required: Mineral Oil (MB1093A), Oxidase Strips (MB0266A), Microbact Software Program (MB1244A), Indole Reagent (MB0209A), VPI Reagent (MB0181A), VPII Reagent (MB0184A), TDA Reagent (MB0180A), NIT A Reagent (MB0186A), NIT B Reagent (MB0187A). The six reagents above are also available as Reagent Set D (MB1082A). 
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
General References:
  1. Farmer, J.J. et al. (1985) J. Clin. Micro. 21(1): 46-76
  2. Burke, V., Robinson, J., Atkinson, H.M. and Gracey, M. (Jan 1982) J. Clin. Micro. 48-52
  3. Cowen, S.T. and Steel, K.J. (1977) Manual for the Identification of Medical Bacteria, 2nd edition, Cambridge University Press.
  4. Balows, A., Hausler, W.J., Herrmann, K.L., Isengerg, J.D. and Shadomy, H.J. (eds) (1991) Manual of Clinical Microbiology, 5th edition, American Society of Microbiology, Washington
Specifications
DescriptionMicrobact GNB 12B Kit
Packaging Type1 Box
Quantity60 Tests/Kit
Detectable AnalytesGram-negative bacilli
Unit SizeEach
Intended use statement 
A qualitative micromethod using enzyme reactions for the identification of aerobic and facultatively anaerobic Gram-negative bacteria (Enterobacteriaceae and miscellaneous Gram- negative bacteria) based on the utilisation of biochemical substrates. Used in a diagnostic workflow to aid clinicians in treatment options for patients suspected of having bacterial infections. The device is not automated, is for professional use only and is not a companion diagnostic.
Microbact users: 
Download Thermo Scientific™ Microbact™ Software and stay up-to-date with our latest updates by visiting our Biochemical Identification Software Downloads portal ›
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Lot #Certificate TypeDateCatalog Number(s)
4502224Certificate of AnalysisMar 29, 2025MB1133A
4486801Certificate of AnalysisMar 25, 2025MB1133A
4488141Certificate of AnalysisMar 25, 2025MB1133A
4490023Certificate of AnalysisMar 25, 2025MB1132A
4487542Certificate of AnalysisMar 25, 2025MB1132A
5 results displayed, search above for a specific certificate
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection