Oxoid™ Microbact™ Staph Fast Blue Reagent Set
Oxoid™ Microbact™ Staph Fast Blue Reagent Set
Thermo Scientific™
Oxoid™ Microbact™ Staph Fast Blue Reagent Set
Simplify the identification of staphylococci, including staphylococcus aureus and coagulase-negative staphylococci, with the Thermo Scientific™ Oxoid™ Microbact™ Staph Fast Blue Reagent Set.
Have Questions?
Catalog NumberDescriptionQuantity
MB1588AMicrobact Staph Fast Blue Reagent Set2 x 10mL Bottles
Catalog number MB1588A
Price (HKD)
-
Request A Quote
Description:
Microbact Staph Fast Blue Reagent Set
Quantity:
2 x 10mL Bottles
Ask our AI about this Product
Simplify the identification of staphylococci, including staphylococcus aureus and coagulase-negative staphylococci (CNS), with the Thermo Scientific™ Oxoid™ Microbact™ Staph Fast Blue Reagent Set. This set is used in the Beta-galactosidase test for fast, reliable and easy-to-read results.
Used in the Beta-galactosidase test, this reagent set allows for quick, reliable and easy identification of staphylococci, including staphylococcus aureus and coagulase-negative staphylococci (CNS).
Not all products are available for sale in all territories. Please inquire.
Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
DescriptionMicrobact Staph Fast Blue Reagent Set
Packaging Type1 Box
Quantity2 x 10mL Bottles
Target Organism ClassStaphylococcus aureus
TypeReagent Set for Staphylococcus aureus Test Kit
Detectable AnalytesStaphylococcus species
Unit SizeEach
Intended use statement 
The Microbact™ Staph Fast Blue Reagent is used with the Microbact™ Staph 12S kit and is based on the detection of Beta-galactosidase.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link