Sensititre™ Gram Negative RUO Susceptibility Testing Plate
Sensititre™ Gram Negative  RUO Susceptibility Testing Plate
Thermo Scientific™
Sensititre™ Gram Negative RUO Susceptibility Testing Plate
NEW antimicrobial options including cefiderocol and imipenem/relebactam together with colistin and fosfomycin for difficult to treat Gram-negative infections in a high-quality, ready to use AST broth microdilution (BMD) plate format, meeting the latest CLSI breakpoints.
Have Questions?
Catalog number MDRGNX2F
Price (USD)
-
Request A Quote
Ask our AI about this Product
Expand your Gram-negative organism susceptibility testing options by accessing the latest antibiotics including cefiderocol and imipenem/relebactam. The Thermo Scientific™ Sensititre™ MDRGNX2F plate delivers results directly from broth microdilution (BMD), the gold standard1 phenotypic method for Antimicrobial Susceptibility Testing (AST). Open access to one of the most comprehensive range of second-line antimicrobials, together with colistin and fosfomycin on a single plate format, designed for research use only.
  • Updated standard format including cefiderocol, imipenem/relebactam, colistin and fosfomycin
  • Research Use Only (RUO)
  • BMD provides accurate MIC results therefore negating the need for repeat testing
  • Designed for use with CLSI breakpoints
  • Sensititre MIC determination is achieved using direct measurement rather than relying on extrapolated data, thereby providing an accurate MIC result, not an estimate.
  • All contained antibiotics also available in customizable plate options, along with access to over 300 antimicrobials, to enable meeting local formulary and Stewardship program requirements
Thermo Fisher Scientific is a leader in providing AST solutions that help in the fight against antimicrobial resistance, offering high performance automated and manual test options and one of the largest, most up-to-date selections of antimicrobials for susceptibility testing.
The Sensititre System utilizes gold standard-level1 MIC accuracy, providing superior quality and reproducibility for accurate results the first time. Broth microdilution is the reference method for AST, and in many cases2 the most reliable phenotypic method to guide therapeutic decisions.
For Research Use Only. Not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Antimicrobial AgentsVarious
DescriptionSensititre Gram Negative RUO AST Plate 10/Pk
Antibiotic CodeVarious
Product TypeAST Plate
Target Organism ClassMultidrug-resistant, Gram-negative Organisms
Test TypeMinimum Inhibitory Concentration (MIC)
Quantity10/Pk.
Unit SizePack of 10
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link