MEP-300 Upgrade Package for MSA and AnStat
MEP-300 Upgrade Package for MSA and AnStat
Thermo Scientific™
MEP-300 Upgrade Package for MSA and AnStat
The Thermo Scientific™ MEP-300 Multi-Element Probe improves XRF elemental slurry analysis without liquid nitrogen.
Have Questions?
Catalog NumberType
MEP300UGMEP-300 Upgrade Package for MSA and AnStat
Catalog number MEP300UG
Price (USD)
-
Request A Quote
Type:
MEP-300 Upgrade Package for MSA and AnStat
Ask our AI about this Product
Improve XRF elemental slurry analysis without the need for liquid nitrogen with the Thermo Scientific™ MEP-300 Multi-Element Probe. The MEP-300 probe uses an advanced Silicon Drift Detector (SDD) and has been developed to replace both MEP and MEP-200 liquid nitrogen cooled X-ray Fluorescence immersion probes used in the AnStat and MSA X-ray analyzer products.
The MEP-300 probe uses an advanced Silicon Drift Detector (SDD) which does not require cryogenic temperatures. The detector resolution and counting rates are improved compared with the previous generation MEP probes. In addition to their excellent X-ray detection capabilities, SDDs have demonstrated superior stability and resistance to shock and vibration in service, making them well suited for industrial applications such as on-line slurry analysis. 
Key design features and advancements:
Occupational Health & Safety
  • No liquid nitrogen required
  • Eliminate Occupational Health & Safety risks previously associated with liquid nitrogen
  • Improved radiation safety 
    • Automatically deployed retractable source with source shield in event of window rupture or when probe raised for servicing
    • Fail safe spring loaded design to withdraw source from window area upon loss of power
    • High visibility beacon for visual status and alarm conditions
Silicon Drift Detector (SDD)
  • Lower detection limits and improved analytical results due to:
    • Higher count rates
    • Lower background
    • Wider field of view
  • Higher availability
  • No special handling or storage requirements 
  • Field repairable - eliminates “returns” to the factory
Maintenance and Operations
  • No financial cost of liquid nitrogen
  • Lower cost of critical spares
  • Field Replaceable modules and cable assemblies
Specifications
Air RequirementsInstrument Air: ANSI ISA-S7 0 0-1996 (clean and dry to 0.1 microns with dew point <2°C)
Air Pressure: 550 kPa nominal - 1000 kPa maximum
Certifications/ComplianceCE, ICES 001, CSA, FCC Part 15B and RCM AS/NZS CISPR11
Electrical RequirementsThree Phase 48 to 62 Hz. Max 2.5kVA Neutral not required. Identical power requirements to previous generation products. Any of the standard three phase voltages used in the world mining industry in the range 380-600 Volts AC may be factory or field selected. Variation should not exceed ±10% of the nominal voltage selected.
Operating TemperatureAmbient -10°C to +55°C (measured at enclosure surface temperature, temperature to not exceed 55°C) Process Fluid: 0°C to 45°C, Check with factory for higher temperature options
TypeMEP-300 Upgrade Package for MSA and AnStat
Unit SizeEach
ramsey-flex-pod-image-278x123
Smart new Thinking for Belt Scale Weighing Systems
Find out more about the Ramsey Flex integration solution!
Learn more ›
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link
1x1 image pixel for data collection