Metallurgical Sample Holder
Metallurgical Sample Holder
Thermo Scientific™
Metallurgical Sample Holder
The base of this sample holder is the same as that of the Standard Sample Holder, except that this holder is designed to support resin-mounted samples. This holder is the preferred solution for metallurgy and when working with inserts.
Have Questions?
Catalog number METALSAMPLEHOLD
Price (USD)
-
Request A Quote
Ask our AI about this Product
The base of this Thermo Scientific™ sample holder is the same as that of the Standard Sample Holder, except that this holder is designed to support resin-mounted samples. This holder is the preferred solution for metallurgy and when working with inserts.
Specifications
  • Sample size up to 32 mm diameter and 30 mm height
Specifications
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link