Neon™ Transfection System 10 μL Kit
The Neon Transfection instrument (Cat. No. MPK5000 and MPK5000S) has been discontinued. The Neon consumables (Cat. No. MPK1025, MPK10025, MPK1096, MPK10096, and MPT100) will be discontinued on September 30, 2025 or while supplies last. Learn about the benefits of the new Neon NxT Electroporation System with improved usability, more protocols with plate setup, and connectivity.
Neon™ Transfection System 10 μL Kit
Invitrogen™

Neon™ Transfection System 10 μL Kit

The Neon™ Transfection System 10 μL Kit is designed specifically for use with the Neon™ Transfection System. Use this kitRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
MPK109696 x 2
MPK102525 x 2
Catalog number MPK1096
Price (USD)/ 96 x 2 reactions
2,106.65
Online exclusive
2235.00 
Save 128.35 (6%)
-
Add to cart
No. of Reactions:
96 x 2
Price (USD)/ 96 x 2 reactions
2,106.65
Online exclusive
2235.00 
Save 128.35 (6%)
Add to cart
Neon™ Transfection System 10 μL Kit
Catalog numberMPK1096
Price (USD)/ 96 x 2 reactions
2,106.65
Online exclusive
2235.00 
Save 128.35 (6%)
-
Add to cart
The Neon™ Transfection System 10 μL Kit is designed specifically for use with the Neon™ Transfection System. Use this kit for transfection volumes of 10 μL, containing 5 × 104–2 × 105 adherent cells or 1 × 105–5 × 105 suspension cells. Cells that have been transfected using the included 10 μL Neon™ Tips are ready to be washed and plated into a multi-well culture dish.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ClassificationAnimal Origin-Free, Chemically Defined, Low-Protein, Xeno-Free, Reduced Growth Factors, Reduced Serum
Cell TypeStem Cells, Primary Cells, Hard-to-Transfect Cells
Format6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample TypePlasmid DNA, Synthetic siRNA
Transfection TechniqueElectroporation
Concentration10 x 10^4 - 2 x 10^5 cells/mL (Adherent Cells), 1 x 10^6 - 5 x 10^5 cells/mL
For Use With (Equipment)Neon™ Transfection System
For Use With (Application)Transfection
No. of Reactions96 x 2
Product LineNeon™
Product TypeTransfection System
Quantity96 x 2 Reactions
Serum CompatibleYes
Shipping ConditionRoom Temperature
Starting Material Amount2x10^4 - 6 x10^8 cells; Varies by cell type
System TypeNeon™ Transfection System
Volume (Metric) Sample10μl, 10 μl
Unit Size96 x 2 reactions
Contents & Storage
• 3 x 1 mL Resuspension Buffer R
• 3 x 1 mL Resuspension Buffer T
• 2 x 150 mL Electrolytic Buffer E
• 96 x 10 μl Neon™ tips
• 20 Neon™ electroporation tubes

Store the buffer box 4°C and the tips and tubes box at room temperature.