Informational:Informational:The Neon Transfection instrument (Cat. No. MPK5000 and MPK5000S) has been discontinued. The Neon consumables (Cat. No. MPK1025, MPK10025, MPK1096, MPK10096, and MPT100) will be discontinued on September 30, 2025 or while supplies last. Learn about the benefits of the new Neon NxT Electroporation System with improved usability, more protocols with plate setup, and connectivity.
Invitrogen™

Neon™ Transfection Tubes

Catalog number: MPT100
Have Questions?
Invitrogen™

Neon™ Transfection Tubes

Catalog number: MPT100
Have Questions?
Catalog Number
MPT100
Unit Size
1 pack
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
MPT1001 pack
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Neon™ Transfection Tubes are designed specifically for use with the Neon™ Transfection System device (MPK5000). Each Neon™ Transfection System Kit (MPK1096 and MPK10096) already comes with 20 electroporation tubes. The additional pack of 100 tubes is offered for those labs wishing to use single tubes per reaction depending on their experimental set-up.

 

For more information on the product, please click here

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample Type
RNA, DNA
For Use With (Equipment)
Neon™ Transfection System
For Use With (Application)
Transfection
Product Line
Neon™
Product Type
Transfection Tube
Quantity
1 Pack
Serum Compatible
Yes
Shipping Condition
Room Temperature
Starting Material Amount
2x10^4 - 6 x10^8 cells; Varies by cell type
System Type
Neon™ Transfection System

Contents & Storage

The Neon™ Transfection Tubes comes in a pack of 100 tubes. Refer to the Neon™ Transfection System users manual for protocol instructions. For use at room temperature under normal laboratory conditions.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text