Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Obtain the minimum inhibitory concentration (MIC) results to first- and second-line anti-tuberculosis drugs for Mycobacterium tuberculosis isolates with Thermo Scientific™ Sensititre™ Mycobacterium tuberculosis MYCOTB AST Plate.
The convenient plate includes 12 antimicrobials on a single AST plate for cost-effective results and is intended for research use only. Easy set-up procedures and incubation requirements provide MIC results in 7 to 21 days, and inclusive on-scale QC ranges provide immediate quality assurance of testing methodology
0.5 McFarland Standard | Saline Tween w/ Glass Beads (Part No. T3490) |
Species | Mycobacterium tuberculosis |
Antimicrobial Agents | Various |
Broth Type | Middlebrook 7H9 w/ OADC (Part No. T3440) |
Description | Sensititre Mycobacterium tuberculosis MIC Plate (U.S.A. only) |
Antibiotic Code | Various |
Product Type | MIC Plate |
Test Type | Minimum Inhibitory Concentration (MIC) |
For Use With (Application) | Mycobacterium tuberculosis |
Incubation Conditions | 35-37°C O2 |
Incubation Time | 10-21 days |
Isolates Per Plate | 1 |
No. per Pack | 10 |
Packaging Type | Box |
Quantity | 10/Pk. |
Reading Options | Manual |
Shelf Life | 18 to 24 Months |
Transfer Volume | 100µL |
Volume Per Well | 100 μL |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Quantity | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
MYCOTB | Each | Sensititre Mycobacterium tuberculosis MIC Plate (U.S.A. only) | 10/Pk. | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Perform comprehensive susceptibility testing of Mycobacterium tuberculosis to first and second line anti-tuberculosis drugs with Sensititre Mycobacterium tuberculosis MYCOTB AST Plate.
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.