Specifications
Product LineArrayScan
For Use With (Equipment)ArrayScan HCA Readers
FormatSlide(s), 96-well plate, 6-well plate, 24-well plate, 8 wells, 384-well plate
OpticsHardware-based Zeiss Definite Focus autofocus system
Software CompatibilityThermo Scientific™ HCS Studio Cell Analysis Software
Operating EnvironmentPermissible ambient temperature range during operation : 18°C to 25°C, Permissible relative humidity during operation (non-condensing): >75% at 30°C
DimensionsControl unit: 22inD x 13in W x 14inH
Weight (Metric)20 kg
QuantityEach
Voltage100-240 VAC
Wattage600 W
Unit Size