Invitrogen™

Neon™ NxT Electroporation System 100-μL Kit

Catalog number: N10025
Have Questions?
Invitrogen™

Neon™ NxT Electroporation System 100-μL Kit

Catalog number: N10025
Have Questions?
Catalog Number
N10025
Unit Size
25 x 2 reactions
Price (USD)
Quantity
Catalog Number
Unit Size
96 x 2 reactions
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
N1002525 x 2 reactions
N1009696 x 2 reactions
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Neon NxT Electroporation System 100-μL Kit is designed specifically for use with the Neon NxT Electroporation System. Use this kit for transfection volumes of 100 μL, containing 5 × 105–2 × 106 adherent cells or 1 × 106–5 × 106 suspension cells. Cells that have been transfected using the included 100-μL Neon NxT tips are then ready to be washed and plated into a multi-well culture dish.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type
Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format
Electroporation Pipette Tip
Transfection Technique
Electroporation
For Use With (Equipment)
Neon™ NxT Electroporation System
For Use With (Application)
Transfection
No. of Reactions
25 x 2 Reactions
Product Type
Pipette Tip
Volume (Metric) Sample
100 μL

Contents & Storage

• 10 mL Resuspension R Buffer; store at 4°C
• 10 mL Resuspension T Buffer; store at 4°C
• 10 mL Resuspension Genome Editing Buffer; store at 4°C
• 50 mL Electrolytic E100 Buffer; store at 4°C
• 25 reaction tips
• 8 electroporation tubes

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text