NBD C6-Ceramide (6-((N-(7-Nitrobenz-2-Oxa-1,3-Diazol-4-yl)amino)hexanoyl)Sphingosine)
NBD C<sub>6</sub>-Ceramide (6-((<i>N</i>-(7-Nitrobenz-2-Oxa-1,3-Diazol-4-yl)amino)hexanoyl)Sphingosine)
Invitrogen™

NBD C6-Ceramide (6-((N-(7-Nitrobenz-2-Oxa-1,3-Diazol-4-yl)amino)hexanoyl)Sphingosine)

NBD C6-ceramide can be used to study sphingolipid transport and metabolism mechanisms. It is also used as a selective stainRead more
Have Questions?
Catalog NumberQuantity
N11541 mg
Catalog number N1154
Price (USD)
404.00
Each
-
Add to cart
Quantity:
1 mg
Price (USD)
404.00
Each
Add to cart
Ask our AI about this Product
NBD C6-ceramide can be used to study sphingolipid transport and metabolism mechanisms. It is also used as a selective stain for the Golgi apparatus in live and fixed cells. The environment sensitive NBD dye is weakly fluorescent in water but increases in aprotic solvents and other nonpolar environments with excitation/emission maxima ∼466/536 nm.

Visualize staining your cell without wasting your reagents, antibodies, or time with our new Stain-iT Cell Staining Simulator.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialSphingolipids
Recommended StorageStore in freezer (-5 to -30°C) and protect from light.
Physical FormSolid
Product LineInvitrogen
Quantity1 mg
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Fluorescence spectra

Fluorescence spectra

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link