TMB Substrate Solution
Product Image
Thermo Scientific™

TMB Substrate Solution

Addition of sulfuric acid stop solution changes the color to yellow, enabling accurate measurement of the intensity at 450nm usingRead more
Have Questions?
Catalog NumberQuantity
N301100 mL
Catalog number N301
Price (KRW)
176,000
Online offer
Ends: 30-Jun-2025
195,000
Save 19,000 (10%)
Each
-
Add to cart
Quantity:
100 mL
Recurring order eligible. Learn more »
Request bulk or custom format
Price (KRW)
176,000
Online offer
Ends: 30-Jun-2025
195,000
Save 19,000 (10%)
Each
Add to cart
Ask our AI about this Product
Addition of sulfuric acid stop solution changes the color to yellow, enabling accurate measurement of the intensity at 450nm using a spectrophotometer or plate reader. This particular formulation is used in most of the Thermo Scientific Pierce ELISA Kits where it provides consistent performance for the sandwich ELISA technique.

Features of TMB Substrate Solution:

Product Number: N301
Size: 100 mL (sufficient for about ten 96-well microplates)
Storage: Shipped on cold packs. Store at 2-8°C immediately upon receipt. Do not use product beyond the stated expiration date.
Application: ELISA: Recommended for peroxidase-based enzyme immunoassays. Upon oxidation, TMB forms a water-soluble blue reaction product that can be measured spectrophotometrically at 605nm. Upon acidification, the reaction product becomes yellow with an absorbance peak at 450nm.
Precautions: This product is sensitive to contamination from a variety of oxidizing agents. To avoid contamination and premature expiration, avoid contacting TMB solution with any potential source of contamination. Never pipette directly from the bottle. Pour out required amount onto a tube and pipette from the tube. Do not return excess TMB to primary storage container.

Procedure:
• Add 100 μL of TMB substrate to each well containing peroxidase reaction.
• Incubate at room temperature for 15 to 30 minutes.
• Add 100 μL of stop solution (0.16M sulfuric acid, N600) to each well.
• Measure absorbance at 450nm.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity100 mL
Substrate PropertiesChemical Substrate
Substrate TypeHRP (Horseradish Peroxidase) Substrate
Detection MethodColorimetric
Product LinePierce™
Product TypeTMB Substrate
Unit SizeEach
Contents & Storage
Store between 2°C and 8°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
ZJ397380Certificate of AnalysisApr 24, 2025N301
AC406348ACertificate of AnalysisApr 15, 2025N301
AC406348Certificate of AnalysisApr 15, 2025N301
ZG395392Certificate of AnalysisAug 17, 2024N301
ZA385019Certificate of AnalysisApr 30, 2024N301
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection