M13 Forward (-20) Primer
M13 Forward (-20) Primer
Invitrogen™

M13 Forward (-20) Primer

Oligonucleotides complementary to a DNA template are necessary to prime DNA synthesis for sequencing reactions. We offer a wide selectionRead more
Have Questions?
Catalog NumberQuantity
N520022 μg
Catalog number N52002
Price (USD)
378.65
Online Exclusive
405.00
Save 26.35 (7%)
Each
Add to cart
Quantity:
2 μg
Price (USD)
378.65
Online Exclusive
405.00
Save 26.35 (7%)
Each
Add to cart

Oligonucleotides complementary to a DNA template are necessary to prime DNA synthesis for sequencing reactions. We offer a wide selection of Invitrogen single-strand primers that may be used in either single- or double-stranded sequencing protocols. All sequencing primers are non-phosphorylated and are supplied lyophilized.

M13 Forward (-20) Primer features include:
• Desalted and purified by gel filtration
• Assayed for function in automated sequencing reactions
• Supplied in quantity of 2 μg

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Sequencing Reactions
FormLyophilized
Primer Length16-mer
Primer Sequence5 ́d[GTAAAACGACGGCCAG]3 ́
Product TypePrimer
Purification MethodDesalted, Gel Filtration Purified
Quantity2 μg
Shipping ConditionRoom Temperature
PrimerM13
Unit SizeEach
Contents & Storage
• M13 Forward (-20) Primer (2 μg)

Store in Freezer at –20°C.
Guaranteed stable for 6 months when properly stored.

Share catalog number, name or link