Informational:Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
Applied Biosystems™

MicroAmp™ 12-Cap Strip

Catalog number: N8011534
Have Questions?
Applied Biosystems™

MicroAmp™ 12-Cap Strip

Catalog number: N8011534
Have Questions?
Catalog Number
N8011534
Unit Size
1,000 strips
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
200 strips
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
N80115341,000 strips
-
N8010534200 strips
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
MicroAmp™ Cap Strips are designed to fit on MicroAmp™ reaction tubes, tube strips, and 96-well plates. They are available in 8-cap and 12-cap strips in clear or an assorted set of red, orange, blue, and green. They have a domed profile and are suitable for end-point PCR. Attach caps using the MicroAmp™ Cap Installing Tool.

Will not interfere with sample reading

• Provides tight seal to minimize evaporation
• Easy to apply and remove

Minimizes Evaporation
Use MicroAmp™ Caps Strips to seal the tubes containing your precious samples. This will reduce the possibility of contamination among sample wells and help ensure reliable results.

Easy to Use
Use the MicroAmp™ Cap Installing Tool to push the caps down and form a tight seal. When ready, just peel away the caps using the tabs at the end.

Colored for Easy Organization
MicroAmp™ Caps Strips are available in different colors to help you keep track of your tubes during assay preparation.

Note: See user's manual or package insert for limited label license and trademark information.

Specifications

Certifications/Compliance
Certified DNA/Rnase Free
Color
Assorted
Quantity
1000 Strips (5 pk./200)
For Use With (Equipment)
Thermal Cyclers
Product Line
MicroAmp™ 
Material
Polypropylene
Closure Type
Domed Caps
No. of Wells
96
Format
1,000 Strip Caps
Product Type
12-Strip Cap

Contents & Storage

Contains 1,000 MicroAmp™ 12-Cap Strips (5 packs of SKU# N8010534, Qty 200 per pack). Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text