Search Thermo Fisher Scientific
Catalog Number | Unit Size | Price (USD) | Availability | Quantity | |
---|---|---|---|---|---|
N8080128 | 5 nmol | 107.00 | - |
Invitrogen Oligo(dT)16 Primer is a homo-oligomeric deoxyribonucleotide poly(dT) that is non-phosphorylated and chemically synthesized. It is used for priming and reverse transcription of polyadenylated poly(A)+ mRNA.
Five nmoles of Oligo (dT)16 Primer is supplied in 100 μL of 10 mM TE, pH 7.0, at a concentration of 50 μM.